Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00544790_m1
          See other SPRED1 GE Assays ›
          Gene Symbol:
          SPRED1
          Gene Name:
          sprouty related EVH1 domain containing 1
          Gene Aliases:
          NFLS, PPP1R147, hSpred1, spred-1
          Chromosome Location:
          Chr.15: 38252087 - 38357249 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00544790_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SPRED1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_152594.2 NP_689807.1 3 - 4 715 95
          XM_005254202.3 XP_005254259.1 4 - 5 1515 95
          XM_011521289.2 XP_011519591.1 3 - 4 412 95
          GenBank mRNA AK091222.1 - 4 - 5 485 95
          AY299089.1 - 3 - 4 715 95
          BC018015.1 - 3 - 4 711 95
          BC137480.1 - 3 - 4 1240 9021480 95
          BC137481.1 - 3 - 4 1240 9021481 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1500006896.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC15000259.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SPRED1

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          inactivation of MAPK activity
          multicellular organism development
          fibroblast growth factor receptor signaling pathway
          negative regulation of peptidyl-threonine phosphorylation
          negative regulation of phosphatase activity
          positive regulation of DNA damage response, signal transduction by p53 class mediator
          regulation of protein deacetylation
          stem cell factor receptor binding
          protein binding
          protein kinase binding
          phosphatase binding
          protein serine/threonine kinase inhibitor activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR15001623.hg.1 ENST00000299084.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15001623.hg.1 NM_152594.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15001623.hg.1 TC15000259.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR15001621.hg.1 ENST00000299084.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15001621.hg.1 ENST00000561205.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15001621.hg.1 NM_152594.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15001621.hg.1 TC15000259.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1500006896.hg.1 ENST00000299084.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006896.hg.1 ENST00000299084.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006896.hg.1 ENST00000299084.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006896.hg.1 ENST00000299084.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006896.hg.1 ENST00000299084.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006896.hg.1 ENST00000299084.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006896.hg.1 ENST00000299084.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006896.hg.1 ENST00000299084.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006896.hg.1 ENST00000299084.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006896.hg.1 ENST00000299084.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006896.hg.1 ENST00000299084.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006896.hg.1 ENST00000299084.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006896.hg.1 ENST00000561205.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006896.hg.1 ENST00000561205.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006896.hg.1 ENST00000561205.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006896.hg.1 ENST00000561205.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006896.hg.1 ENST00000561205.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006896.hg.1 ENST00000561205.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006896.hg.1 ENST00000561317.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006896.hg.1 ENST00000561317.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006896.hg.1 ENST00000561317.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006896.hg.1 ENST00000561317.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006896.hg.1 ENST00000561317.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006896.hg.1 ENST00000561317.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006896.hg.1 NM_152594.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006896.hg.1 NM_152594.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006896.hg.1 NM_152594.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006896.hg.1 NM_152594.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006896.hg.1 NM_152594.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006896.hg.1 NM_152594.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006896.hg.1 NM_152594.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006896.hg.1 NM_152594.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006896.hg.1 NM_152594.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006896.hg.1 NM_152594.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006896.hg.1 NM_152594.2 Refseq
          Affymetrix Human Clariom D Assay PSR1500147790.hg.1 ENST00000299084.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1500147790.hg.1 ENST00000561205.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500147790.hg.1 ENST00000561317.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500147790.hg.1 NM_152594.2 Refseq
          Affymetrix Human Clariom D Assay PSR1500147790.hg.1 ENST00000299084.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1500147790.hg.1 ENST00000561205.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500147790.hg.1 ENST00000561317.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500147790.hg.1 NM_152594.2 Refseq
          Affymetrix Human Clariom D Assay PSR1500147790.hg.1 TC1500006896.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500147793.hg.1 ENST00000299084.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1500147793.hg.1 NM_152594.2 Refseq
          Affymetrix Human Clariom D Assay PSR1500147793.hg.1 ENST00000299084.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1500147793.hg.1 NM_152594.2 Refseq
          Affymetrix Human Clariom D Assay PSR1500147793.hg.1 TC1500006896.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1500006896.hg.1 ENST00000299084.8 Ensembl
          Affymetrix Human Clariom D Assay TC1500006896.hg.1 ENST00000299084.8 Ensembl
          Affymetrix Human Clariom D Assay TC1500006896.hg.1 ENST00000299084.8 Ensembl
          Affymetrix Human Clariom D Assay TC1500006896.hg.1 ENST00000299084.8 Ensembl
          Affymetrix Human Clariom D Assay TC1500006896.hg.1 ENST00000299084.8 Ensembl
          Affymetrix Human Clariom D Assay TC1500006896.hg.1 ENST00000299084.8 Ensembl
          Affymetrix Human Clariom D Assay TC1500006896.hg.1 ENST00000299084.8 Ensembl
          Affymetrix Human Clariom D Assay TC1500006896.hg.1 ENST00000299084.8 Ensembl
          Affymetrix Human Clariom D Assay TC1500006896.hg.1 ENST00000299084.8 Ensembl
          Affymetrix Human Clariom D Assay TC1500006896.hg.1 ENST00000299084.8 Ensembl
          Affymetrix Human Clariom D Assay TC1500006896.hg.1 ENST00000299084.8 Ensembl
          Affymetrix Human Clariom D Assay TC1500006896.hg.1 ENST00000299084.8 Ensembl
          Affymetrix Human Clariom D Assay TC1500006896.hg.1 ENST00000561205.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006896.hg.1 ENST00000561205.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006896.hg.1 ENST00000561205.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006896.hg.1 ENST00000561205.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006896.hg.1 ENST00000561205.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006896.hg.1 ENST00000561205.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006896.hg.1 ENST00000561317.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006896.hg.1 ENST00000561317.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006896.hg.1 ENST00000561317.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006896.hg.1 ENST00000561317.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006896.hg.1 ENST00000561317.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006896.hg.1 ENST00000561317.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006896.hg.1 NM_152594.2 Refseq
          Affymetrix Human Clariom D Assay TC1500006896.hg.1 NM_152594.2 Refseq
          Affymetrix Human Clariom D Assay TC1500006896.hg.1 NM_152594.2 Refseq
          Affymetrix Human Clariom D Assay TC1500006896.hg.1 NM_152594.2 Refseq
          Affymetrix Human Clariom D Assay TC1500006896.hg.1 NM_152594.2 Refseq
          Affymetrix Human Clariom D Assay TC1500006896.hg.1 NM_152594.2 Refseq
          Affymetrix Human Clariom D Assay TC1500006896.hg.1 NM_152594.2 Refseq
          Affymetrix Human Clariom D Assay TC1500006896.hg.1 NM_152594.2 Refseq
          Affymetrix Human Clariom D Assay TC1500006896.hg.1 NM_152594.2 Refseq
          Affymetrix Human Clariom D Assay TC1500006896.hg.1 NM_152594.2 Refseq
          Affymetrix Human Clariom D Assay TC1500006896.hg.1 NM_152594.2 Refseq
          Affymetrix Human Clariom D Assay PSR1500147792.hg.1 ENST00000299084.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1500147792.hg.1 ENST00000561317.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500147792.hg.1 NM_152594.2 Refseq
          Affymetrix Human Clariom D Assay PSR1500147792.hg.1 ENST00000299084.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1500147792.hg.1 ENST00000561317.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500147792.hg.1 NM_152594.2 Refseq
          Affymetrix Human Clariom D Assay PSR1500147792.hg.1 TC1500006896.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 223349 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline