| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13005775.hg.1 |
ENST00000316334.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13005775.hg.1 |
NM_153371.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13005775.hg.1 |
TC13000511.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008443.hg.1 |
ENST00000316334.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008443.hg.1 |
ENST00000316334.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008443.hg.1 |
ENST00000316334.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008443.hg.1 |
ENST00000316334.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008443.hg.1 |
ENST00000316334.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008443.hg.1 |
ENST00000316334.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008443.hg.1 |
ENST00000316334.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008443.hg.1 |
ENST00000316334.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008443.hg.1 |
ENST00000316334.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008443.hg.1 |
ENST00000316334.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008443.hg.1 |
ENST00000316334.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008443.hg.1 |
ENST00000316334.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008443.hg.1 |
NM_153371.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008443.hg.1 |
NM_153371.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008443.hg.1 |
NM_153371.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008443.hg.1 |
NM_153371.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008443.hg.1 |
NM_153371.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008443.hg.1 |
NM_153371.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008443.hg.1 |
NM_153371.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008443.hg.1 |
NM_153371.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008443.hg.1 |
NM_153371.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008443.hg.1 |
NM_153371.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008443.hg.1 |
NM_153371.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008443.hg.1 |
NM_153371.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008443.hg.1 |
NM_153371.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008443.hg.1 |
ENST00000316334.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008443.hg.1 |
ENST00000316334.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008443.hg.1 |
ENST00000316334.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008443.hg.1 |
ENST00000316334.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008443.hg.1 |
ENST00000316334.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008443.hg.1 |
ENST00000316334.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008443.hg.1 |
ENST00000316334.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008443.hg.1 |
ENST00000316334.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008443.hg.1 |
ENST00000316334.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008443.hg.1 |
ENST00000316334.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008443.hg.1 |
ENST00000316334.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008443.hg.1 |
ENST00000316334.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008443.hg.1 |
NM_153371.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008443.hg.1 |
NM_153371.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008443.hg.1 |
NM_153371.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008443.hg.1 |
NM_153371.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008443.hg.1 |
NM_153371.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008443.hg.1 |
NM_153371.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008443.hg.1 |
NM_153371.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008443.hg.1 |
NM_153371.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008443.hg.1 |
NM_153371.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008443.hg.1 |
NM_153371.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008443.hg.1 |
NM_153371.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008443.hg.1 |
NM_153371.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008443.hg.1 |
NM_153371.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300156918.hg.1 |
ENST00000316334.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300156918.hg.1 |
NM_153371.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300156918.hg.1 |
ENST00000316334.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300156918.hg.1 |
NM_153371.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300156918.hg.1 |
TC1300008443.hg.1 |
Clariom D TC |