Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00745341_s1
          See other OLIG1 GE Assays ›
          Gene Symbol:
          OLIG1
          Gene Name:
          oligodendrocyte transcription factor 1
          Gene Aliases:
          BHLHB6, BHLHE21
          Chromosome Location:
          Chr.21: 33070144 - 33072422 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00745341_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: OLIG1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138983.2 NP_620450.2 1 148 118
          GenBank mRNA AB209791.1 - 1 139 118
          AL834450.1 - 1 141 118
          BC033290.1 - 1 155 4940343 118

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2100006951.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC21000125.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AL834450.1,BC033290.1,NM_138983.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene OLIG1

          Panther Classification:

          Molecular Function -

          basic helix-loop-helix transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          neuron fate commitment
          oligodendrocyte differentiation
          DNA binding
          protein binding
          protein dimerization activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC2100006951.hg.1 ENST00000382348.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006951.hg.1 ENST00000382348.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006951.hg.1 ENST00000382348.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006951.hg.1 ENST00000382348.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006951.hg.1 ENST00000382348.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006951.hg.1 ENST00000382348.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006951.hg.1 ENST00000382348.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006951.hg.1 ENST00000426947.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006951.hg.1 ENST00000426947.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006951.hg.1 ENST00000426947.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006951.hg.1 ENST00000498799.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006951.hg.1 NM_138983.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006951.hg.1 NM_138983.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006951.hg.1 NM_138983.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006951.hg.1 NM_138983.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006951.hg.1 NM_138983.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006951.hg.1 NM_138983.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006951.hg.1 NM_138983.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR21000822.hg.1 ENST00000333063.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21000822.hg.1 ENST00000382348.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21000822.hg.1 NM_138983.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR21000822.hg.1 TC21000125.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR21000823.hg.1 ENST00000333063.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21000823.hg.1 ENST00000382348.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21000823.hg.1 NM_138983.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR21000823.hg.1 TC21000125.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR2100146638.hg.1 ENST00000382348.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146638.hg.1 NM_138983.2 Refseq
          Affymetrix Human Clariom D Assay PSR2100146638.hg.1 TC2100006951.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100146638.hg.1 ENST00000382348.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146638.hg.1 NM_138983.2 Refseq
          Affymetrix Human Clariom D Assay TC2100006951.hg.1 ENST00000382348.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100006951.hg.1 ENST00000382348.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100006951.hg.1 ENST00000382348.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100006951.hg.1 ENST00000382348.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100006951.hg.1 ENST00000382348.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100006951.hg.1 ENST00000382348.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100006951.hg.1 ENST00000382348.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100006951.hg.1 ENST00000426947.5 Ensembl
          Affymetrix Human Clariom D Assay TC2100006951.hg.1 ENST00000426947.5 Ensembl
          Affymetrix Human Clariom D Assay TC2100006951.hg.1 ENST00000426947.5 Ensembl
          Affymetrix Human Clariom D Assay TC2100006951.hg.1 ENST00000498799.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100006951.hg.1 NM_138983.2 Refseq
          Affymetrix Human Clariom D Assay TC2100006951.hg.1 NM_138983.2 Refseq
          Affymetrix Human Clariom D Assay TC2100006951.hg.1 NM_138983.2 Refseq
          Affymetrix Human Clariom D Assay TC2100006951.hg.1 NM_138983.2 Refseq
          Affymetrix Human Clariom D Assay TC2100006951.hg.1 NM_138983.2 Refseq
          Affymetrix Human Clariom D Assay TC2100006951.hg.1 NM_138983.2 Refseq
          Affymetrix Human Clariom D Assay TC2100006951.hg.1 NM_138983.2 Refseq
          Affymetrix Human Clariom D Assay PSR2100146636.hg.1 ENST00000382348.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146636.hg.1 NM_138983.2 Refseq
          Affymetrix Human Clariom D Assay PSR2100146636.hg.1 TC2100006951.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100146636.hg.1 ENST00000382348.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146636.hg.1 NM_138983.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 185472 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline