Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00904548_m1
          See other ALG2 GE Assays ›
          Gene Symbol:
          ALG2
          Gene Name:
          ALG2, alpha-1,3/1,6-mannosyltransferase
          Gene Aliases:
          CDGIi, CMS14, CMSTA3, NET38, hALPG2
          Chromosome Location:
          Chr.9: 99216425 - 99221964 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00904548_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ALG2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_024532.1 2 - 3 578 150
          GenBank mRNA AK074988.1 - 2 - 3 556 150
          AY358697.1 - 1 - 2 163 150
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AI768004.1 - 2 - 3 221 Chr. 9 - 99218594 - 99221607 [-] on Build GRCh38 150
          GenBankmRNA AW051481.1 - 2 - 3 221 Chr. 9 - 99218621 - 99221607 [-] on Build GRCh38 150
          GenBankmRNA BP342204.1 - 2 - 3 526 Chr. 9 - 99218778 - 99221912 [-] on Build GRCh38 150
          GenBankmRNA CF132606.1 - 2 - 3 345 Chr. 9 - 99218712 - 99221731 [-] on Build GRCh38 150
          GenBankmRNA CF131378.1 - 2 - 3 377 Chr. 9 - 99218633 - 99221763 [-] on Build GRCh38 150
          GenBankmRNA DA430466.1 - 2 - 3 527 Chr. 9 - 99218796 - 99221912 [-] on Build GRCh38 150
          GenBankmRNA DA736744.1 - 2 - 3 556 Chr. 9 - 99218649 - 99221942 [-] on Build GRCh38 150
          GenBankmRNA DR006890.1 - 2 - 3 435 Chr. 9 - 99218507 - 99221816 [-] on Build GRCh38 150
          GenBankmRNA AI863884.1 - 2 - 3 221 Chr. 9 - 99218654 - 99221607 [-] on Build GRCh38 150
          GenBankmRNA BQ641538.1 - 468 Chr. - - [-] on Build 150
          GenBankmRNA BU594798.1 - 2 - 3 559 Chr. 9 - 99218625 - 99221944 [-] on Build GRCh38 150
          GenBankmRNA BG773528.1 - 2 - 3 373 Chr. 9 - 99218494 - 99221523 [-] on Build GRCh38 150
          GenBankmRNA HY019575.1 - 2 - 3 288 Chr. 9 - 99218635 - 99221548 [-] on Build GRCh38 150
          GenBankmRNA AA769683.1 - 2 - 3 220 Chr. 9 - 99218552 - 99221606 [-] on Build GRCh38 150
          GenBankmRNA DB187551.1 - 2 - 3 531 Chr. 9 - 99218789 - 99221912 [-] on Build GRCh38 150
          GenBankmRNA AI278375.1 - 2 - 3 221 Chr. 9 - 99218609 - 99221607 [-] on Build GRCh38 150
          GenBankmRNA DA714128.1 - 2 - 3 536 Chr. 9 - 99218795 - 99221914 [-] on Build GRCh38 150
          GenBankmRNA AI183323.1 - 2 - 3 221 Chr. 9 - 99218659 - 99221607 [-] on Build GRCh38 150
          GenBankmRNA CB046477.1 - 1 - 2 Chr. 9 - 99218381 - 99221140 [+] on Build GRCh38 150

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream transforming growth factor beta receptor 1 TGFBR1 AAT5,
          ACVRLK4,
          ALK-5,
          ALK5,
          ESS1,
          LDS1,
          LDS1A,
          LDS2A,
          MSSE,
          SKR4,
          TGFR-1,
          tbetaR-I
          -64.4kb Anti-sense Chr. 9 - 99104038 - 99154192 on NCBI Build GRCh38
          Downstream Sec61 translocon beta subunit SEC61B 1.2kb Anti-sense Chr. 9 - 99222288 - 99230619 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0900010995.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC09001411.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK074988.1,AY358697.1 transcripts.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          dolichol-linked oligosaccharide biosynthetic process
          protein glycosylation in endoplasmic reticulum
          response to calcium ion
          mannosylation
          alpha-1,3-mannosyltransferase activity
          GDP-Man:Man1GlcNAc2-PP-Dol alpha-1,3-mannosyltransferase activity
          protein binding
          protein heterodimerization activity
          protein N-terminus binding
          calcium-dependent protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0900010995.hg.1 ENST00000238477.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010995.hg.1 ENST00000238477.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010995.hg.1 ENST00000238477.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010995.hg.1 ENST00000238477.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010995.hg.1 ENST00000238477.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010995.hg.1 ENST00000238477.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010995.hg.1 ENST00000319033.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010995.hg.1 ENST00000319033.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010995.hg.1 ENST00000319033.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010995.hg.1 ENST00000476832.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010995.hg.1 ENST00000476832.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010995.hg.1 ENST00000476832.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010995.hg.1 ENST00000476832.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010995.hg.1 NM_033087.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010995.hg.1 NM_033087.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010995.hg.1 NM_033087.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010995.hg.1 NM_033087.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010995.hg.1 NM_033087.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010995.hg.1 NR_024532.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010995.hg.1 NR_024532.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010995.hg.1 NR_024532.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010995.hg.1 NR_024532.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010995.hg.1 NR_024532.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010995.hg.1 NR_024532.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09017598.hg.1 NM_033087.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09017598.hg.1 NR_024532.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09017598.hg.1 TC09001411.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR09017602.hg.1 ENST00000319033.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09017602.hg.1 NR_024532.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09017602.hg.1 TC09001411.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0900010995.hg.1 ENST00000238477.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010995.hg.1 ENST00000238477.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010995.hg.1 ENST00000238477.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010995.hg.1 ENST00000238477.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010995.hg.1 ENST00000238477.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010995.hg.1 ENST00000238477.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900010995.hg.1 ENST00000319033.7 Ensembl
          Affymetrix Human Clariom D Assay TC0900010995.hg.1 ENST00000319033.7 Ensembl
          Affymetrix Human Clariom D Assay TC0900010995.hg.1 ENST00000319033.7 Ensembl
          Affymetrix Human Clariom D Assay TC0900010995.hg.1 ENST00000476832.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900010995.hg.1 ENST00000476832.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900010995.hg.1 ENST00000476832.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900010995.hg.1 ENST00000476832.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900010995.hg.1 NM_033087.3 Refseq
          Affymetrix Human Clariom D Assay TC0900010995.hg.1 NM_033087.3 Refseq
          Affymetrix Human Clariom D Assay TC0900010995.hg.1 NM_033087.3 Refseq
          Affymetrix Human Clariom D Assay TC0900010995.hg.1 NM_033087.3 Refseq
          Affymetrix Human Clariom D Assay TC0900010995.hg.1 NM_033087.3 Refseq
          Affymetrix Human Clariom D Assay TC0900010995.hg.1 NR_024532.1 Refseq
          Affymetrix Human Clariom D Assay TC0900010995.hg.1 NR_024532.1 Refseq
          Affymetrix Human Clariom D Assay TC0900010995.hg.1 NR_024532.1 Refseq
          Affymetrix Human Clariom D Assay TC0900010995.hg.1 NR_024532.1 Refseq
          Affymetrix Human Clariom D Assay TC0900010995.hg.1 NR_024532.1 Refseq
          Affymetrix Human Clariom D Assay TC0900010995.hg.1 NR_024532.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900174936.hg.1 ENST00000238477.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174936.hg.1 NM_033087.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900174936.hg.1 NR_024532.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900174936.hg.1 ENST00000238477.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174936.hg.1 NM_033087.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900174936.hg.1 NR_024532.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900174936.hg.1 TC0900010995.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900174939.hg.1 ENST00000238477.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174939.hg.1 ENST00000319033.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174939.hg.1 ENST00000476832.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174939.hg.1 NM_033087.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900174939.hg.1 NR_024532.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900174939.hg.1 ENST00000238477.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174939.hg.1 ENST00000319033.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174939.hg.1 ENST00000476832.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174939.hg.1 NM_033087.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900174939.hg.1 NR_024532.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900174939.hg.1 TC0900010995.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline