Search
Search
| Gene Symbol: LYST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000081.3 | NP_000072.2 | 22 - 23 | 6440 | 72 | |
| NM_001301365.1 | NP_001288294.1 | 22 - 23 | 6877 | 72 | ||
| XM_011544031.1 | XP_011542333.1 | 22 - 23 | 6440 | 72 | ||
| XM_011544032.1 | XP_011542334.1 | 22 - 23 | 6843 | 72 | ||
| XM_011544033.2 | XP_011542335.1 | 22 - 23 | 6590 | 72 | ||
| XM_011544034.1 | XP_011542336.1 | 21 - 22 | 6302 | 72 | ||
| XM_011544035.2 | XP_011542337.1 | 22 - 23 | 6440 | 72 | ||
| XM_011544036.2 | XP_011542338.1 | 18 - 19 | 4094 | 72 | ||
| XM_011544037.2 | XP_011542339.1 | 22 - 23 | 6440 | 72 | ||
| XM_011544039.2 | XP_011542341.1 | 22 - 23 | 6440 | 72 | ||
| XM_017000150.1 | XP_016855639.1 | 22 - 23 | 6440 | 72 | ||
| GenBank mRNA | BC144702.1 | - | 21 - 22 | 6314 | 72 | |
| U67615.1 | - | 22 - 23 | 6447 | 72 | ||
| U70064.1 | - | 22 - 23 | 6650 | 72 | ||
Set Membership: |
|