Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00929113_m1
          See other CYSLTR1 GE Assays ›
          Gene Symbol:
          CYSLTR1
          Gene Name:
          cysteinyl leukotriene receptor 1
          Gene Aliases:
          CYSLT1, CYSLT1R, CYSLTR, HMTMF81
          Chromosome Location:
          Chr.X: 78271468 - 78327691 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00929113_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CYSLTR1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001282187.1 NP_001269116.1 3 - 4 560 128
          NM_001282188.1 NP_001269117.1 3 - 4 537 128
          NM_006639.3 NP_006630.1 2 - 3 477 128
          GenBank mRNA AK313643.1 - 2 - 3 401 128
          BC035750.1 - 2 - 3 269 5750221 128
          DQ131799.1 - 2 - 3 477 128
          DQ131801.1 - 3 - 4 404 128
          DQ131802.1 - 3 - 4 411 128

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK313643.1, BC035750.1, DQ131799.1, DQ131801.1, DQ131802.1, NM_001282187.1, NM_001282188.1, NM_006639.3 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CYSLTR1

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          circulatory system process
          calcium ion transport
          chemotaxis
          defense response
          cell surface receptor signaling pathway
          phospholipase C-activating G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          neuropeptide signaling pathway
          respiratory gaseous exchange
          leukotriene signaling pathway
          cysteinyl leukotriene receptor activity
          leukotriene receptor activity
          protein binding
          G-protein coupled peptide receptor activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 ENST00000373304.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 ENST00000373304.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 ENST00000373304.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 ENST00000373304.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 ENST00000373304.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 ENST00000373304.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 ENST00000493254.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 ENST00000493254.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 ENST00000493254.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 ENST00000614798.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 ENST00000614798.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 ENST00000614798.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 ENST00000614798.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 ENST00000614798.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 ENST00000614798.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 ENST00000614798.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 NM_001282186.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 NM_001282186.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 NM_001282186.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 NM_001282186.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 NM_001282186.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 NM_001282186.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 NM_001282186.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 NM_001282187.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 NM_001282187.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 NM_001282187.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 NM_001282187.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 NM_001282187.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 NM_001282187.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 NM_001282187.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 NM_001282187.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 NM_001282187.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 NM_001282188.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 NM_001282188.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 NM_001282188.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 NM_001282188.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 NM_001282188.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 NM_001282188.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 NM_001282188.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 NM_001282188.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 NM_001282188.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 NM_006639.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 NM_006639.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 NM_006639.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 NM_006639.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 NM_006639.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 NM_006639.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 NM_006639.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00010141.hg.1 NM_006639.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 ENST00000373304.3 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 ENST00000373304.3 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 ENST00000373304.3 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 ENST00000373304.3 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 ENST00000373304.3 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 ENST00000373304.3 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 ENST00000493254.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 ENST00000493254.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 ENST00000493254.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 ENST00000614798.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 ENST00000614798.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 ENST00000614798.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 ENST00000614798.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 ENST00000614798.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 ENST00000614798.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 ENST00000614798.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 NM_001282186.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 NM_001282186.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 NM_001282186.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 NM_001282186.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 NM_001282186.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 NM_001282186.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 NM_001282186.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 NM_001282187.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 NM_001282187.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 NM_001282187.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 NM_001282187.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 NM_001282187.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 NM_001282187.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 NM_001282187.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 NM_001282187.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 NM_001282187.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 NM_001282188.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 NM_001282188.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 NM_001282188.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 NM_001282188.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 NM_001282188.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 NM_001282188.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 NM_001282188.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 NM_001282188.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 NM_001282188.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 NM_006639.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 NM_006639.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 NM_006639.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 NM_006639.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 NM_006639.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 NM_006639.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 NM_006639.3 Refseq
          Affymetrix Human Clariom D Assay TC0X00010141.hg.1 NM_006639.3 Refseq
          Affymetrix Human Clariom D Assay PSR0X00169238.hg.1 ENST00000373304.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00169238.hg.1 ENST00000614798.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00169238.hg.1 NM_001282186.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00169238.hg.1 NM_001282187.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00169238.hg.1 NM_001282188.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00169238.hg.1 NM_006639.3 Refseq
          Affymetrix Human Clariom D Assay PSR0X00169238.hg.1 ENST00000373304.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00169238.hg.1 ENST00000614798.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00169238.hg.1 NM_001282186.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00169238.hg.1 NM_001282187.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00169238.hg.1 NM_001282188.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00169238.hg.1 NM_006639.3 Refseq
          Affymetrix Human Clariom D Assay PSR0X00169238.hg.1 TC0X00010141.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0X00169241.hg.1 NM_001282188.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00169241.hg.1 NM_001282188.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00169241.hg.1 TC0X00010141.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0X00169237.hg.1 ENST00000373304.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00169237.hg.1 ENST00000614798.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00169237.hg.1 NM_001282186.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00169237.hg.1 NM_001282187.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00169237.hg.1 NM_001282188.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00169237.hg.1 NM_006639.3 Refseq
          Affymetrix Human Clariom D Assay PSR0X00169237.hg.1 ENST00000373304.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00169237.hg.1 ENST00000614798.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00169237.hg.1 NM_001282186.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00169237.hg.1 NM_001282187.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00169237.hg.1 NM_001282188.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00169237.hg.1 NM_006639.3 Refseq
          Affymetrix Human Clariom D Assay PSR0X00169237.hg.1 TC0X00010141.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0X00169240.hg.1 NM_001282187.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00169240.hg.1 NM_001282187.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00169240.hg.1 TC0X00010141.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline