Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00932200_g1
          See other GTF2A2 GE Assays ›
          Gene Symbol:
          GTF2A2
          Gene Name:
          general transcription factor IIA subunit 2
          Gene Aliases:
          HsT18745, T18745, TF2A2, TFIIA, TFIIA-12, TFIIA-gamma, TFIIAS
          Chromosome Location:
          Chr.15: 59638062 - 59657541 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00932200_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: GTF2A2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001320929.1 NP_001307858.1 3 - 4 358 77
          NM_001320930.1 NP_001307859.1 4 - 5 582 77
          NM_004492.2 NP_004483.1 3 - 4 360 77
          GenBank mRNA AK312015.1 - 3 - 4 360 77
          BC000287.2 - 3 - 4 322 2819215 77
          BC001919.1 - 3 - 4 322 2819215 77
          CB156109.1 - 4 - 5 524 77
          CR542192.1 - 2 - 3 178 77
          HY002503.1 - 3 - 4 345 77
          U14193.1 - 3 - 4 319 77
          U21242.1 - 3 - 4 288 77
          X81713.1 - 3 - 4 277 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1500009621.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC15001501.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene GTF2A2

          Panther Classification:

          Molecular Function -

          general transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription from RNA polymerase II promoter
          transcription initiation from RNA polymerase II promoter
          transcription elongation from RNA polymerase II promoter
          viral process
          snRNA transcription from RNA polymerase II promoter
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of sequence-specific DNA binding transcription factor activity
          RNA polymerase II transcriptional preinitiation complex assembly
          RNA polymerase II repressing transcription factor binding
          transcription coactivator activity
          protein binding
          transcription factor binding
          TBP-class protein binding
          protein homodimerization activity
          protein heterodimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000267869.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000267869.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000267869.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000267869.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000267869.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000396060.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000396060.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000396060.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000396060.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000396060.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000396060.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000396060.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000396060.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000396060.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000396061.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000396061.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000396061.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000396061.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000396061.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000396061.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000396061.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000396061.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000396063.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000396063.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000396063.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000396063.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000396063.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000396063.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000396063.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000396063.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000396063.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000396064.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000396064.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000396064.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000396064.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000396064.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000396064.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000472508.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000472508.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000472508.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000480768.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000480768.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000480768.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000480768.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000484743.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000484743.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000484743.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000484743.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000484743.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000484743.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000559141.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000559141.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000559141.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000559141.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000559706.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000559706.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000559706.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000559706.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 ENST00000559706.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 NM_004492.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 NM_004492.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 NM_004492.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 NM_004492.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 NM_004492.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 NM_004492.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 NM_004492.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 NM_004492.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009621.hg.1 NM_004492.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15014138.hg.1 ENST00000267869.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15014138.hg.1 ENST00000396060.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15014138.hg.1 ENST00000396061.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15014138.hg.1 ENST00000396063.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15014138.hg.1 ENST00000484743.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15014138.hg.1 NM_004492.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15014138.hg.1 TC15001501.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1500170899.hg.1 ENST00000267869.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1500170899.hg.1 ENST00000396060.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1500170899.hg.1 ENST00000396061.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500170899.hg.1 ENST00000396063.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500170899.hg.1 ENST00000484743.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500170899.hg.1 ENST00000559141.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500170899.hg.1 ENST00000559706.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500170899.hg.1 NM_004492.2 Refseq
          Affymetrix Human Clariom D Assay PSR1500170899.hg.1 TC1500009621.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500170899.hg.1 ENST00000267869.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1500170899.hg.1 ENST00000396060.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1500170899.hg.1 ENST00000396061.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500170899.hg.1 ENST00000396063.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500170899.hg.1 ENST00000484743.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500170899.hg.1 ENST00000559141.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500170899.hg.1 ENST00000559706.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500170899.hg.1 NM_004492.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000267869.8 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000267869.8 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000267869.8 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000267869.8 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000267869.8 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000396060.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000396060.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000396060.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000396060.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000396060.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000396060.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000396060.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000396060.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000396060.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000396061.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000396061.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000396061.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000396061.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000396061.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000396061.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000396061.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000396061.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000396063.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000396063.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000396063.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000396063.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000396063.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000396063.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000396063.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000396063.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000396063.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000396064.7 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000396064.7 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000396064.7 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000396064.7 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000396064.7 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000396064.7 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000472508.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000472508.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000472508.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000480768.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000480768.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000480768.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000480768.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000484743.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000484743.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000484743.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000484743.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000484743.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000484743.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000559141.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000559141.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000559141.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000559141.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000559706.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000559706.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000559706.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000559706.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 ENST00000559706.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 NM_004492.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 NM_004492.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 NM_004492.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 NM_004492.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 NM_004492.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 NM_004492.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 NM_004492.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 NM_004492.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009621.hg.1 NM_004492.2 Refseq
          Affymetrix Human Clariom D Assay PSR1500170898.hg.1 ENST00000267869.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1500170898.hg.1 ENST00000396060.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1500170898.hg.1 ENST00000396061.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500170898.hg.1 ENST00000396063.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500170898.hg.1 ENST00000484743.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500170898.hg.1 ENST00000559141.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500170898.hg.1 NM_004492.2 Refseq
          Affymetrix Human Clariom D Assay PSR1500170898.hg.1 TC1500009621.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500170898.hg.1 ENST00000267869.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1500170898.hg.1 ENST00000396060.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1500170898.hg.1 ENST00000396061.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500170898.hg.1 ENST00000396063.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500170898.hg.1 ENST00000484743.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500170898.hg.1 ENST00000559141.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500170898.hg.1 NM_004492.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline