Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00964538_g1
          See other STK38 GE Assays ›
          Gene Symbol:
          STK38
          Gene Name:
          serine/threonine kinase 38
          Gene Aliases:
          NDR, NDR1
          Chromosome Location:
          Chr.6: 36493882 - 36547516 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00964538_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: STK38
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001305102.1 NP_001292031.1 11 - 12 1408 69
          NM_007271.3 NP_009202.1 11 - 12 1411 69
          XM_006714988.3 XP_006715051.1 11 - 12 1791 69
          XM_006714989.3 XP_006715052.1 11 - 12 1788 69
          XM_017010226.1 XP_016865715.1 11 - 12 1371 69
          GenBank mRNA AK095337.1 - 1 - 2 1298 69
          AK312559.1 - 11 - 12 1319 69
          BC012085.1 - 11 - 12 1365 4554884 69
          BC095413.1 - 11 - 12 1371 30337276 69
          Z35102.1 - 10 - 11 1674 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600011661.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06001675.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene STK38

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular protein modification process
          protein phosphorylation
          peptidyl-serine phosphorylation
          intracellular signal transduction
          negative regulation of MAP kinase activity
          cell-cell adhesion
          magnesium ion binding
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          mitogen-activated protein kinase kinase kinase binding
          cadherin binding involved in cell-cell adhesion

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1600007194.hg.1 ENST00000566764.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007194.hg.1 ENST00000566764.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007194.hg.1 ENST00000566764.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 ENST00000229812.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 ENST00000229812.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 ENST00000229812.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 ENST00000229812.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 ENST00000229812.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 ENST00000229812.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 ENST00000229812.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 ENST00000229812.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 ENST00000229812.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 ENST00000229812.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 ENST00000229812.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 ENST00000229812.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 ENST00000229812.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 ENST00000229812.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 ENST00000229812.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 ENST00000229812.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 NM_001305102.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 NM_001305102.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 NM_001305102.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 NM_001305102.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 NM_001305102.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 NM_001305102.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 NM_001305102.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 NM_001305102.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 NM_001305102.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 NM_001305102.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 NM_001305102.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 NM_001305102.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 NM_001305102.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 NM_001305102.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 NM_001305102.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 NM_001305102.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 NM_001305102.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 NM_007271.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 NM_007271.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 NM_007271.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 NM_007271.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 NM_007271.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 NM_007271.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 NM_007271.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 NM_007271.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 NM_007271.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 NM_007271.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 NM_007271.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 NM_007271.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 NM_007271.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 NM_007271.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 NM_007271.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 NM_007271.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011661.hg.1 NM_007271.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06019951.hg.1 ENST00000229812.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06019951.hg.1 NM_007271.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06019951.hg.1 TC06001675.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 ENST00000229812.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 ENST00000229812.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 ENST00000229812.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 ENST00000229812.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 ENST00000229812.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 ENST00000229812.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 ENST00000229812.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 ENST00000229812.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 ENST00000229812.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 ENST00000229812.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 ENST00000229812.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 ENST00000229812.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 ENST00000229812.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 ENST00000229812.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 ENST00000229812.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 ENST00000229812.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 NM_001305102.1 Refseq
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 NM_001305102.1 Refseq
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 NM_001305102.1 Refseq
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 NM_001305102.1 Refseq
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 NM_001305102.1 Refseq
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 NM_001305102.1 Refseq
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 NM_001305102.1 Refseq
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 NM_001305102.1 Refseq
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 NM_001305102.1 Refseq
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 NM_001305102.1 Refseq
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 NM_001305102.1 Refseq
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 NM_001305102.1 Refseq
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 NM_001305102.1 Refseq
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 NM_001305102.1 Refseq
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 NM_001305102.1 Refseq
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 NM_001305102.1 Refseq
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 NM_001305102.1 Refseq
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 NM_007271.3 Refseq
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 NM_007271.3 Refseq
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 NM_007271.3 Refseq
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 NM_007271.3 Refseq
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 NM_007271.3 Refseq
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 NM_007271.3 Refseq
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 NM_007271.3 Refseq
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 NM_007271.3 Refseq
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 NM_007271.3 Refseq
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 NM_007271.3 Refseq
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 NM_007271.3 Refseq
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 NM_007271.3 Refseq
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 NM_007271.3 Refseq
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 NM_007271.3 Refseq
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 NM_007271.3 Refseq
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 NM_007271.3 Refseq
          Affymetrix Human Clariom D Assay TC0600011661.hg.1 NM_007271.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600180009.hg.1 ENST00000229812.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0600180009.hg.1 NM_001305102.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600180009.hg.1 NM_007271.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600180009.hg.1 ENST00000229812.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0600180009.hg.1 NM_001305102.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600180009.hg.1 NM_007271.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600180009.hg.1 TC0600011661.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1600007194.hg.1 ENST00000566764.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600007194.hg.1 ENST00000566764.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600007194.hg.1 ENST00000566764.1 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline