Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00972650_m1
          See other TERT GE Assays ›
          Gene Symbol:
          TERT
          Gene Name:
          telomerase reverse transcriptase
          Gene Aliases:
          CMM9, DKCA2, DKCB4, EST2, PFBMFT1, TCS1, TP2, TRT, hEST2, hTRT
          Chromosome Location:
          Chr.5: 1253167 - 1295626 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00972650_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TERT
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001193376.1 NP_001180305.1 3 - 4 1826 57
          NM_198253.2 NP_937983.2 3 - 4 1826 57
          XM_017009796.1 XP_016865285.1 3 - 4 2398 57
          GenBank mRNA AB085628.1 - 3 - 4 1768 57
          AB086379.1 - 3 - 4 1768 57
          AB086950.1 - 3 - 4 1768 57
          AF015950.1 - 3 - 4 1823 57
          AF018167.1 - 3 - 4 1824 57
          AX810036.1 - 3 - 4 1823 57
          AX810378.1 - 3 - 4 1823 57
          BC062321.1 - 1 - 2 581 57
          JF896280.1 - 2 - 3 437 57

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0500009852.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC05001105.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens Human Signal Transduction Pathways Human Telomere Extension by Telomerase Inventoried Human_Telomere_Extension_by_Telomerase Human_Signal_Transduction_Pathways Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TERT

          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          telomere maintenance
          transcription, RNA-templated
          RNA-dependent DNA biosynthetic process
          telomere maintenance via telomerase
          mitochondrion organization
          negative regulation of gene expression
          DNA strand elongation
          positive regulation of Wnt signaling pathway
          production of siRNA involved in RNA interference
          regulation of protein stability
          positive regulation of protein binding
          RNA biosynthetic process
          positive regulation of hair cycle
          positive regulation of nitric-oxide synthase activity
          establishment of protein localization to telomere
          cellular response to hypoxia
          DNA biosynthetic process
          replicative senescence
          positive regulation of pri-miRNA transcription from RNA polymerase II promoter
          negative regulation of production of siRNA involved in RNA interference
          positive regulation of protein localization to nucleolus
          beta-catenin-TCF complex assembly
          positive regulation of stem cell proliferation
          negative regulation of cellular senescence
          negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
          tRNA binding
          transcription coactivator binding
          DNA binding
          telomerase activity
          telomerase RNA reverse transcriptase activity
          RNA binding
          RNA-directed DNA polymerase activity
          RNA-directed RNA polymerase activity
          protein binding
          nucleotidyltransferase activity
          telomeric DNA binding
          protein homodimerization activity
          metal ion binding
          telomerase RNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR05016066.hg.1 ENST00000296820.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05016066.hg.1 ENST00000310581.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05016066.hg.1 ENST00000334602.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05016066.hg.1 ENST00000508104.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05016066.hg.1 NM_001193376.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05016066.hg.1 NM_198253.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05016066.hg.1 TC05001105.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0500169532.hg.1 ENST00000310581.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0500169532.hg.1 ENST00000334602.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0500169532.hg.1 ENST00000460137.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0500169532.hg.1 ENST00000484238.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0500169532.hg.1 ENST00000508104.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500169532.hg.1 NM_001193376.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500169532.hg.1 NM_198253.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500169532.hg.1 ENST00000310581.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0500169532.hg.1 ENST00000334602.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0500169532.hg.1 ENST00000460137.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0500169532.hg.1 ENST00000484238.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0500169532.hg.1 ENST00000508104.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500169532.hg.1 NM_001193376.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500169532.hg.1 NM_198253.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500169532.hg.1 TC0500009852.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-cbn8l:80/100.66.77.38:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline