Search
Search

DYRK1B| Gene Symbol: DYRK1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004714.2 | NP_004705.1 | 8 - 9 | 1405 | 190 | |
| NM_006484.2 | NP_006475.1 | 8 - 9 | 1405 | 106 | ||
| XM_005259398.4 | XP_005259455.1 | 8 - 9 | 1857 | 190 | ||
| GenBank mRNA | AF205861.1 | - | 8 - 9 | 1302 | 190 | |
| AK074971.1 | - | 8 - 9 | 1321 | 190 | ||
| BC018751.1 | - | 8 - 9 | 1357 | 4300012 | 190 | |
| BC025291.1 | - | 8 - 9 | 1308 | 5019417 | 190 | |
| Y17999.1 | - | 8 - 9 | 1373 | 190 | ||
Set Membership: |
|