Search
Search

PLCE1| Gene Symbol: PLCE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001165979.2 | NP_001159451.1 | 26 - 27 | 5144 | 92 | |
| NM_001288989.1 | NP_001275918.1 | 27 - 28 | 6585 | 92 | ||
| NM_016341.3 | NP_057425.3 | 27 - 28 | 6633 | 92 | ||
| XM_006717885.3 | XP_006717948.1 | 28 - 29 | 6623 | 92 | ||
| XM_006717888.3 | XP_006717951.1 | 28 - 29 | 6620 | 92 | ||
| XM_006717889.3 | XP_006717952.1 | 28 - 29 | 6575 | 92 | ||
| XM_006717890.2 | XP_006717953.1 | 27 - 28 | 5139 | 92 | ||
| XM_011539849.2 | XP_011538151.1 | 28 - 29 | 7780 | 92 | ||
| XM_011539850.2 | XP_011538152.1 | 27 - 28 | 6268 | 92 | ||
| XM_017016310.1 | XP_016871799.1 | 29 - 30 | 6670 | 92 | ||
| XM_017016311.1 | XP_016871800.1 | 28 - 29 | 6623 | 92 | ||
| XM_017016312.1 | XP_016871801.1 | 26 - 27 | 5049 | 92 | ||
| GenBank mRNA | AB040949.2 | - | 27 - 28 | 6633 | 92 | |
| AF117948.1 | - | 18 - 19 | 2708 | 92 | ||
| AF170071.1 | - | 26 - 27 | 5075 | 92 | ||
| AF190642.2 | - | 26 - 27 | 6234 | 92 | ||
| AK022543.1 | - | 10 - 11 | 1589 | 92 | ||
| AK074239.1 | - | 6 - 7 | 1020 | 92 | ||
| AY995135.1 | - | 1 - 2 | 43 | 92 | ||
| BC140705.1 | - | 26 - 27 | 6059 | 9021573 | 92 | |
| BC144286.1 | - | 26 - 27 | 6011 | 9052806 | 92 | |
| BC151854.1 | - | 27 - 28 | 6633 | 8860521 | 92 | |
Set Membership: |
|