Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01071292_m1
          See other PAX1 GE Assays ›
          Gene Symbol:
          PAX1
          Gene Name:
          paired box 1
          Gene Aliases:
          HUP48, OFC2
          Chromosome Location:
          Chr.20: 21705659 - 21718486 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01071292_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PAX1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
          RefSeq NM_001257096.1 NP_001244025.1 2 - 3 969 85
          NM_006192.4 NP_006183.2 2 - 3 969 85
          GenBank mRNA AK303335.1 - 1 - 2 850 85
          BC069134.1 - 1 - 2 633 85
          BC143791.1 - 2 - 3 717 85
          BC143793.1 - 2 - 3 714 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2000006913.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC20000145.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PAX1

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal system development
          somitogenesis
          transcription from RNA polymerase II promoter
          cell proliferation
          CD4-positive, alpha-beta T cell differentiation
          CD8-positive, alpha-beta T cell differentiation
          positive regulation of transcription from RNA polymerase II promoter
          thymus development
          parathyroid gland development
          bone morphogenesis
          sclerotome development
          DNA binding
          transcription factor activity, sequence-specific DNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR20001913.hg.1 ENST00000398485.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20001913.hg.1 ENST00000444366.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20001913.hg.1 ENST00000460221.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20001913.hg.1 NM_006192.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20001913.hg.1 TC20000145.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR20001915.hg.1 ENST00000398485.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20001915.hg.1 ENST00000444366.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20001915.hg.1 ENST00000460221.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20001915.hg.1 NM_006192.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20001915.hg.1 TC20000145.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000398485.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000398485.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000398485.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000398485.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000398485.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000398485.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000398485.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000398485.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000398485.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000398485.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000398485.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000398485.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000398485.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000444366.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000444366.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000444366.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000444366.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000444366.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000444366.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000444366.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000444366.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000444366.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000460221.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000460221.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000460221.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000460221.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000460221.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000460221.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000485038.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000485038.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000485038.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000613128.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000613128.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000613128.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000613128.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000613128.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000613128.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000613128.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000613128.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000613128.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000613128.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000613128.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000613128.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000613128.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 ENST00000613128.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 NM_001257096.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 NM_001257096.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 NM_001257096.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 NM_001257096.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 NM_001257096.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 NM_001257096.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 NM_001257096.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 NM_001257096.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 NM_001257096.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 NM_001257096.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 NM_001257096.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 NM_001257096.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 NM_001257096.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 NM_001257096.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 NM_006192.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 NM_006192.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 NM_006192.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 NM_006192.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 NM_006192.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 NM_006192.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 NM_006192.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 NM_006192.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 NM_006192.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 NM_006192.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 NM_006192.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 NM_006192.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 NM_006192.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006913.hg.1 NM_006192.4 Refseq
          Affymetrix Human Clariom D Assay PSR2000147336.hg.1 ENST00000398485.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2000147336.hg.1 ENST00000444366.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000147336.hg.1 ENST00000460221.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000147336.hg.1 ENST00000485038.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000147336.hg.1 ENST00000613128.4 Ensembl
          Affymetrix Human Clariom D Assay PSR2000147336.hg.1 NM_001257096.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000147336.hg.1 NM_006192.4 Refseq
          Affymetrix Human Clariom D Assay PSR2000147336.hg.1 TC2000006913.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000147336.hg.1 ENST00000398485.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2000147336.hg.1 ENST00000444366.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000147336.hg.1 ENST00000460221.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000147336.hg.1 ENST00000485038.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000147336.hg.1 ENST00000613128.4 Ensembl
          Affymetrix Human Clariom D Assay PSR2000147336.hg.1 NM_001257096.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000147336.hg.1 NM_006192.4 Refseq
          Affymetrix Human Clariom D Assay PSR2000147334.hg.1 ENST00000398485.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2000147334.hg.1 ENST00000444366.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000147334.hg.1 ENST00000460221.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000147334.hg.1 ENST00000613128.4 Ensembl
          Affymetrix Human Clariom D Assay PSR2000147334.hg.1 NM_001257096.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000147334.hg.1 NM_006192.4 Refseq
          Affymetrix Human Clariom D Assay PSR2000147334.hg.1 TC2000006913.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000147334.hg.1 ENST00000398485.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2000147334.hg.1 ENST00000444366.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000147334.hg.1 ENST00000460221.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000147334.hg.1 ENST00000613128.4 Ensembl
          Affymetrix Human Clariom D Assay PSR2000147334.hg.1 NM_001257096.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000147334.hg.1 NM_006192.4 Refseq
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000398485.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000398485.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000398485.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000398485.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000398485.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000398485.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000398485.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000398485.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000398485.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000398485.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000398485.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000398485.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000398485.6 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000444366.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000444366.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000444366.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000444366.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000444366.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000444366.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000444366.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000444366.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000444366.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000460221.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000460221.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000460221.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000460221.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000460221.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000460221.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000485038.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000485038.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000485038.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000613128.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000613128.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000613128.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000613128.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000613128.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000613128.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000613128.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000613128.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000613128.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000613128.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000613128.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000613128.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000613128.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 ENST00000613128.4 Ensembl
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 NM_001257096.1 Refseq
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 NM_001257096.1 Refseq
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 NM_001257096.1 Refseq
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 NM_001257096.1 Refseq
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 NM_001257096.1 Refseq
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 NM_001257096.1 Refseq
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 NM_001257096.1 Refseq
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 NM_001257096.1 Refseq
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 NM_001257096.1 Refseq
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 NM_001257096.1 Refseq
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 NM_001257096.1 Refseq
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 NM_001257096.1 Refseq
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 NM_001257096.1 Refseq
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 NM_001257096.1 Refseq
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 NM_006192.4 Refseq
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 NM_006192.4 Refseq
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 NM_006192.4 Refseq
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 NM_006192.4 Refseq
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 NM_006192.4 Refseq
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 NM_006192.4 Refseq
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 NM_006192.4 Refseq
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 NM_006192.4 Refseq
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 NM_006192.4 Refseq
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 NM_006192.4 Refseq
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 NM_006192.4 Refseq
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 NM_006192.4 Refseq
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 NM_006192.4 Refseq
          Affymetrix Human Clariom D Assay TC2000006913.hg.1 NM_006192.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch