Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01085331_m1
          See other ACE2 GE Assays ›
          Gene Symbol:
          ACE2
          Gene Name:
          angiotensin I converting enzyme 2
          Gene Aliases:
          ACEH
          Chromosome Location:
          Chr.X: 15494525 - 15602069 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01085331_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ACE2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
          RefSeq NM_021804.2 NP_068576.1 15 - 16 2114 101
          XM_011545549.1 XP_011543851.1 14 - 15 1951 101
          XM_011545551.2 XP_011543853.1 14 - 15 1951 101
          GenBank mRNA AB193259.1 - 15 - 16 2200 101
          AB193260.1 - 14 - 15 2065 101
          AF241254.1 - 14 - 15 2000 101
          AF291820.1 - 14 - 15 1917 101
          AK315144.1 - 15 - 16 2115 101
          AL110224.1 - 14 - 15 1896 101
          AY623811.1 - 14 - 15 1897 101
          BC039902.1 - 14 - 15 1915 101
          BC048094.1 - 14 - 15 1931 101
          BC048144.1 - 14 - 15 1931 101
          GQ262784.1 - 14 - 15 1897 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0X00009128.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC0X000878.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ACE2

          Panther Classification:

          Molecular Function -

          metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cytokine production
          angiotensin maturation
          angiotensin catabolic process in blood
          angiotensin-mediated drinking behavior
          regulation of systemic arterial blood pressure by renin-angiotensin
          tryptophan transport
          regulation of vasoconstriction
          receptor biosynthetic process
          regulation of cell proliferation
          regulation of vasodilation
          viral entry into host cell
          receptor-mediated virion attachment to host cell
          regulation of inflammatory response
          positive regulation of cardiac muscle contraction
          positive regulation of gap junction assembly
          regulation of cardiac conduction
          positive regulation of reactive oxygen species metabolic process
          virus receptor activity
          glycoprotein binding
          endopeptidase activity
          carboxypeptidase activity
          metallocarboxypeptidase activity
          protein binding
          metallopeptidase activity
          peptidyl-dipeptidase activity
          zinc ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR0X011753.hg.1 ENST00000252519.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X011753.hg.1 ENST00000427411.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X011753.hg.1 NM_021804.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X011753.hg.1 TC0X000878.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0X00162642.hg.1 ENST00000252519.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00162642.hg.1 ENST00000427411.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00162642.hg.1 NM_021804.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00162642.hg.1 ENST00000252519.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00162642.hg.1 ENST00000427411.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00162642.hg.1 NM_021804.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00162642.hg.1 TC0X00009128.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-74676f44b6-vpxpb:80/100.66.72.150:80.
          git-commit: db542085ed6beb0c3f0dd774c2548ab49b9f1b6a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026.10.42-1.0