Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01100785_m1
          See other NOL9 GE Assays ›
          Gene Symbol:
          NOL9
          Gene Name:
          nucleolar protein 9
          Gene Aliases:
          Grc3, NET6
          Chromosome Location:
          Chr.1: 6521347 - 6554598 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01100785_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: NOL9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024654.4 NP_078930.3 9 - 10 1737 68
          XM_005263493.3 XP_005263550.1 7 - 8 1069 68
          XM_011542147.2 XP_011540449.1 4 - 5 663 68
          XM_017002338.1 XP_016857827.1 7 - 8 1247 68
          XM_017002339.1 XP_016857828.1 6 - 7 963 68
          GenBank mRNA AK026976.1 - 5 - 6 747 68
          AK091284.1 - 9 - 10 1674 68
          AK289653.1 - 9 - 10 1669 68
          BC009257.2 - 6 - 7 827 68
          BC012439.1 - 2 - 3 313 68
          BC063639.1 - 6 - 7 777 68
          BC073860.1 - 5 - 6 740 68
          BC094836.1 - 8 - 9 1112 68
          BC105095.1 - 9 - 10 1737 8144098 68
          BC110849.1 - 9 - 10 1646 6167575 68
          BC112278.1 - 9 - 10 1737 8327746 68
          BC143646.1 - 9 - 10 1737 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100012687.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01002154.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene NOL9

          Panther Classification:

          Molecular Function -

          mRNA polyadenylation factor

          Gene Ontology Categories:

          Function(s) Process(es)

          cleavage in ITS2 between 5.8S rRNA and LSU-rRNA of tricistronic rRNA transcript (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
          maturation of 5.8S rRNA
          rRNA processing
          phosphorylation
          RNA binding
          protein binding
          ATP binding
          polynucleotide 5'-hydroxyl-kinase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01033258.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01033258.hg.1 NM_024654.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01033258.hg.1 TC01002154.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 ENST00000460777.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 ENST00000460777.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 ENST00000460777.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 ENST00000460777.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 ENST00000462798.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 ENST00000462798.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 ENST00000462798.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 ENST00000464383.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 ENST00000464383.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 ENST00000464383.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 ENST00000464383.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 ENST00000464383.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 ENST00000464383.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 ENST00000464383.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 ENST00000464665.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 ENST00000464665.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 ENST00000464665.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 NM_024654.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 NM_024654.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 NM_024654.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 NM_024654.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 NM_024654.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 NM_024654.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 NM_024654.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 NM_024654.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 NM_024654.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 NM_024654.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 NM_024654.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 NM_024654.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 NM_024654.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 NM_024654.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 NM_024654.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 NM_024654.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 NM_024654.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 NM_024654.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 NM_024654.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012687.hg.1 NM_024654.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01033260.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01033260.hg.1 ENST00000464383.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01033260.hg.1 NM_024654.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01033260.hg.1 TC01002154.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01033259.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01033259.hg.1 NM_024654.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01033259.hg.1 TC01002154.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0100194019.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100194019.hg.1 ENST00000464383.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100194019.hg.1 NM_024654.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100194019.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100194019.hg.1 ENST00000464383.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100194019.hg.1 NM_024654.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100194019.hg.1 TC0100012687.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100194017.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100194017.hg.1 NM_024654.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100194017.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100194017.hg.1 NM_024654.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100194017.hg.1 TC0100012687.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100194018.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100194018.hg.1 NM_024654.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100194018.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100194018.hg.1 NM_024654.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100194018.hg.1 TC0100012687.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 ENST00000377705.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 ENST00000460777.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 ENST00000460777.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 ENST00000460777.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 ENST00000460777.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 ENST00000462798.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 ENST00000462798.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 ENST00000462798.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 ENST00000464383.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 ENST00000464383.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 ENST00000464383.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 ENST00000464383.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 ENST00000464383.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 ENST00000464383.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 ENST00000464383.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 ENST00000464665.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 ENST00000464665.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 ENST00000464665.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 NM_024654.4 Refseq
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 NM_024654.4 Refseq
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 NM_024654.4 Refseq
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 NM_024654.4 Refseq
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 NM_024654.4 Refseq
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 NM_024654.4 Refseq
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 NM_024654.4 Refseq
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 NM_024654.4 Refseq
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 NM_024654.4 Refseq
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 NM_024654.4 Refseq
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 NM_024654.4 Refseq
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 NM_024654.4 Refseq
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 NM_024654.4 Refseq
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 NM_024654.4 Refseq
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 NM_024654.4 Refseq
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 NM_024654.4 Refseq
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 NM_024654.4 Refseq
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 NM_024654.4 Refseq
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 NM_024654.4 Refseq
          Affymetrix Human Clariom D Assay TC0100012687.hg.1 NM_024654.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline