| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20011282.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20011282.hg.1 |
ENST00000373694.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20011282.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20011282.hg.1 |
TC20000821.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000465985.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000465985.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000465985.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
ENST00000465985.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000465985.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000465985.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000465985.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
ENST00000465985.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009023.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000163211.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000163211.hg.1 |
NM_015474.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000163211.hg.1 |
TC2000009023.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000163211.hg.1 |
ENST00000262878.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000163211.hg.1 |
NM_015474.3 |
Refseq |