Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01124251_g1
          See other CXCL10 GE Assays ›
          Gene Symbol:
          CXCL10
          Gene Name:
          C-X-C motif chemokine ligand 10
          Gene Aliases:
          C7, IFI10, INP10, IP-10, SCYB10, crg-2, gIP-10, mob-1
          Chromosome Location:
          Chr.4: 76021118 - 76023497 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01124251_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CXCL10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001565.4 NP_001556.2 2 - 3 250 135
          GenBank mRNA BC010954.1 - 2 - 3 254 135
          X02530.1 - 2 - 3 250 135

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0400011053.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC04001305.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CXCL10

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of leukocyte chemotaxis
          chemotaxis
          defense response
          inflammatory response
          immune response
          signal transduction
          cell surface receptor signaling pathway
          G-protein coupled receptor signaling pathway
          adenylate cyclase-activating G-protein coupled receptor signaling pathway
          cell-cell signaling
          muscle organ development
          blood circulation
          positive regulation of cell proliferation
          response to bacterium
          response to gamma radiation
          T cell chemotaxis
          regulation of T cell chemotaxis
          response to auditory stimulus
          negative regulation of angiogenesis
          positive regulation of cell migration
          neutrophil chemotaxis
          killing of cells of other organism
          response to lipopolysaccharide
          response to vitamin D
          cellular response to heat
          endothelial cell activation
          regulation of cell proliferation
          regulation of apoptotic process
          negative regulation of myoblast differentiation
          positive regulation of transcription from RNA polymerase II promoter
          positive chemotaxis
          positive regulation of release of sequestered calcium ion into cytosol
          defense response to virus
          antimicrobial humoral immune response mediated by antimicrobial peptide
          chemokine-mediated signaling pathway
          cellular response to lipopolysaccharide
          positive regulation of monocyte chemotaxis
          cellular response to interleukin-17
          cellular response to virus
          antiviral innate immune response
          regulation of endothelial tube morphogenesis
          negative regulation of myoblast fusion
          positive regulation of T cell migration
          receptor binding
          cytokine activity
          protein binding
          chemokine activity
          heparin binding
          cAMP-dependent protein kinase regulator activity
          chemoattractant activity
          CXCR chemokine receptor binding
          CXCR3 chemokine receptor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR04017886.hg.1 ENST00000306602.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04017886.hg.1 NM_001565.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04017886.hg.1 TC04001305.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0400011053.hg.1 ENST00000306602.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011053.hg.1 ENST00000306602.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011053.hg.1 ENST00000306602.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011053.hg.1 ENST00000306602.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011053.hg.1 ENST00000306602.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011053.hg.1 NM_001565.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011053.hg.1 NM_001565.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011053.hg.1 NM_001565.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011053.hg.1 NM_001565.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011053.hg.1 NM_001565.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011053.hg.1 NM_001565.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400175300.hg.1 ENST00000306602.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0400175300.hg.1 NM_001565.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400175300.hg.1 ENST00000306602.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0400175300.hg.1 NM_001565.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400175300.hg.1 TC0400011053.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0400011053.hg.1 ENST00000306602.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400011053.hg.1 ENST00000306602.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400011053.hg.1 ENST00000306602.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400011053.hg.1 ENST00000306602.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400011053.hg.1 ENST00000306602.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400011053.hg.1 NM_001565.3 Refseq
          Affymetrix Human Clariom D Assay TC0400011053.hg.1 NM_001565.3 Refseq
          Affymetrix Human Clariom D Assay TC0400011053.hg.1 NM_001565.3 Refseq
          Affymetrix Human Clariom D Assay TC0400011053.hg.1 NM_001565.3 Refseq
          Affymetrix Human Clariom D Assay TC0400011053.hg.1 NM_001565.3 Refseq
          Affymetrix Human Clariom D Assay TC0400011053.hg.1 NM_001565.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline