Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01572709_g1
          See other PSMB6 GE Assays ›
          Gene Symbol:
          PSMB6
          Gene Name:
          proteasome subunit beta 6
          Gene Aliases:
          DELTA, LMPY, Y
          Chromosome Location:
          Chr.17: 4796144 - 4798503 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01572709_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PSMB6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001270481.1 NP_001257410.1 4 - 5 478 115
          NM_002798.2 NP_002789.1 4 - 5 478 115
          GenBank mRNA AK311768.1 - 4 - 5 441 115
          BC000835.2 - 4 - 5 445 3456009 115
          BM553399.1 - 4 - 5 425 115
          BQ691763.1 - 4 - 5 460 115
          BT009809.1 - 4 - 5 427 115
          BU848746.1 - 4 - 5 410 115
          CR457065.1 - 4 - 5 427 115

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700006644.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17000052.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protease 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PSMB6

          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          protein polyubiquitination
          stimulatory C-type lectin receptor signaling pathway
          antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-dependent
          regulation of cellular amino acid metabolic process
          viral process
          anaphase-promoting complex-dependent catabolic process
          tumor necrosis factor-mediated signaling pathway
          NIK/NF-kappaB signaling
          Fc-epsilon receptor signaling pathway
          proteasome-mediated ubiquitin-dependent protein catabolic process
          regulation of mRNA stability
          T cell receptor signaling pathway
          negative regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
          positive regulation of ubiquitin-protein ligase activity involved in regulation of mitotic cell cycle transition
          Wnt signaling pathway, planar cell polarity pathway
          negative regulation of canonical Wnt signaling pathway
          positive regulation of canonical Wnt signaling pathway
          cell-cell adhesion
          endopeptidase activity
          threonine-type endopeptidase activity
          cadherin binding involved in cell-cell adhesion

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 ENST00000270586.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 ENST00000270586.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 ENST00000270586.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 ENST00000270586.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 ENST00000270586.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 ENST00000270586.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 ENST00000270586.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 ENST00000270586.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 ENST00000270586.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 ENST00000571309.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 ENST00000571309.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 ENST00000571309.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 ENST00000571309.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 ENST00000571309.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 ENST00000571309.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 ENST00000575079.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 ENST00000575079.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 ENST00000575079.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 ENST00000575643.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 ENST00000575643.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 ENST00000575643.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 ENST00000575643.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 ENST00000575643.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 ENST00000575643.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 ENST00000575643.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 ENST00000614486.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 ENST00000614486.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 ENST00000614486.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 ENST00000614486.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 ENST00000614486.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 ENST00000614486.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 ENST00000614486.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 ENST00000614486.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 ENST00000614486.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 NM_001270481.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 NM_001270481.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 NM_001270481.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 NM_001270481.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 NM_001270481.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 NM_001270481.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 NM_001270481.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 NM_001270481.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 NM_001270481.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 NM_002798.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 NM_002798.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 NM_002798.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 NM_002798.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 NM_002798.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 NM_002798.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 NM_002798.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 NM_002798.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006644.hg.1 NM_002798.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17000529.hg.1 ENST00000270586.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17000529.hg.1 NM_002798.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17000529.hg.1 TC17000052.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1700145478.hg.1 ENST00000270586.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1700145478.hg.1 ENST00000571309.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700145478.hg.1 ENST00000575643.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700145478.hg.1 ENST00000614486.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700145478.hg.1 NM_001270481.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700145478.hg.1 NM_002798.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700145478.hg.1 TC1700006644.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700145478.hg.1 ENST00000270586.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1700145478.hg.1 ENST00000571309.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700145478.hg.1 ENST00000575643.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700145478.hg.1 ENST00000614486.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700145478.hg.1 NM_001270481.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700145478.hg.1 NM_002798.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700145476.hg.1 ENST00000270586.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1700145476.hg.1 ENST00000571309.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700145476.hg.1 ENST00000575643.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700145476.hg.1 ENST00000614486.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700145476.hg.1 NM_001270481.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700145476.hg.1 NM_002798.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700145476.hg.1 TC1700006644.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700145476.hg.1 ENST00000270586.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1700145476.hg.1 ENST00000571309.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700145476.hg.1 ENST00000575643.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700145476.hg.1 ENST00000614486.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700145476.hg.1 NM_001270481.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700145476.hg.1 NM_002798.2 Refseq
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 ENST00000270586.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 ENST00000270586.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 ENST00000270586.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 ENST00000270586.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 ENST00000270586.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 ENST00000270586.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 ENST00000270586.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 ENST00000270586.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 ENST00000270586.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 ENST00000571309.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 ENST00000571309.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 ENST00000571309.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 ENST00000571309.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 ENST00000571309.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 ENST00000571309.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 ENST00000575079.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 ENST00000575079.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 ENST00000575079.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 ENST00000575643.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 ENST00000575643.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 ENST00000575643.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 ENST00000575643.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 ENST00000575643.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 ENST00000575643.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 ENST00000575643.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 ENST00000614486.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 ENST00000614486.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 ENST00000614486.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 ENST00000614486.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 ENST00000614486.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 ENST00000614486.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 ENST00000614486.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 ENST00000614486.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 ENST00000614486.4 Ensembl
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 NM_001270481.1 Refseq
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 NM_001270481.1 Refseq
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 NM_001270481.1 Refseq
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 NM_001270481.1 Refseq
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 NM_001270481.1 Refseq
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 NM_001270481.1 Refseq
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 NM_001270481.1 Refseq
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 NM_001270481.1 Refseq
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 NM_001270481.1 Refseq
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 NM_002798.2 Refseq
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 NM_002798.2 Refseq
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 NM_002798.2 Refseq
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 NM_002798.2 Refseq
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 NM_002798.2 Refseq
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 NM_002798.2 Refseq
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 NM_002798.2 Refseq
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 NM_002798.2 Refseq
          Affymetrix Human Clariom D Assay TC1700006644.hg.1 NM_002798.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700145475.hg.1 ENST00000270586.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1700145475.hg.1 ENST00000575643.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700145475.hg.1 ENST00000614486.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700145475.hg.1 NM_001270481.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700145475.hg.1 NM_002798.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700145475.hg.1 TC1700006644.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700145475.hg.1 ENST00000270586.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1700145475.hg.1 ENST00000575643.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700145475.hg.1 ENST00000614486.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1700145475.hg.1 NM_001270481.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700145475.hg.1 NM_002798.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline