| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004737.hg.1 |
ENST00000297001.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004737.hg.1 |
NM_001161834.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004737.hg.1 |
TC07000312.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004736.hg.1 |
ENST00000297001.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004736.hg.1 |
NM_001161834.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004736.hg.1 |
TC07000312.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007501.hg.1 |
ENST00000297001.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007501.hg.1 |
ENST00000297001.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007501.hg.1 |
ENST00000297001.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007501.hg.1 |
ENST00000297001.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007501.hg.1 |
ENST00000297001.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007501.hg.1 |
ENST00000297001.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007501.hg.1 |
ENST00000297001.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007501.hg.1 |
ENST00000297001.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007501.hg.1 |
ENST00000297001.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007501.hg.1 |
ENST00000297001.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007501.hg.1 |
ENST00000297001.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007501.hg.1 |
ENST00000297001.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007501.hg.1 |
NM_001161834.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007501.hg.1 |
NM_001161834.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007501.hg.1 |
NM_001161834.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007501.hg.1 |
NM_001161834.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007501.hg.1 |
NM_001161834.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007501.hg.1 |
NM_001161834.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007501.hg.1 |
NM_001161834.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007501.hg.1 |
NM_001161834.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007501.hg.1 |
NM_001161834.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007501.hg.1 |
NM_001161834.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700152168.hg.1 |
ENST00000297001.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700152168.hg.1 |
NM_001161834.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700152168.hg.1 |
ENST00000297001.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700152168.hg.1 |
NM_001161834.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700152168.hg.1 |
TC0700007501.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700152167.hg.1 |
ENST00000297001.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700152167.hg.1 |
NM_001161834.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700152167.hg.1 |
ENST00000297001.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700152167.hg.1 |
NM_001161834.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700152167.hg.1 |
TC0700007501.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0700007501.hg.1 |
ENST00000297001.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007501.hg.1 |
ENST00000297001.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007501.hg.1 |
ENST00000297001.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007501.hg.1 |
ENST00000297001.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007501.hg.1 |
ENST00000297001.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007501.hg.1 |
ENST00000297001.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007501.hg.1 |
ENST00000297001.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007501.hg.1 |
ENST00000297001.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007501.hg.1 |
ENST00000297001.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007501.hg.1 |
ENST00000297001.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007501.hg.1 |
ENST00000297001.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007501.hg.1 |
ENST00000297001.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007501.hg.1 |
NM_001161834.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007501.hg.1 |
NM_001161834.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007501.hg.1 |
NM_001161834.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007501.hg.1 |
NM_001161834.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007501.hg.1 |
NM_001161834.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007501.hg.1 |
NM_001161834.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007501.hg.1 |
NM_001161834.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007501.hg.1 |
NM_001161834.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007501.hg.1 |
NM_001161834.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007501.hg.1 |
NM_001161834.2 |
Refseq |