Search
Search

SLC16A7| Gene Symbol: SLC16A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270622.1 | NP_001257551.1 | 3 - 4 | 518 | 77 | |
| NM_001270623.1 | NP_001257552.1 | 3 - 4 | 538 | 77 | ||
| NM_004731.4 | NP_004722.2 | 2 - 3 | 376 | 77 | ||
| XM_005269231.4 | XP_005269288.1 | 2 - 3 | 312 | 77 | ||
| GenBank mRNA | AF049608.1 | - | 3 - 4 | 373 | 77 | |
| AF058056.1 | - | 2 - 3 | 339 | 77 | ||
| AK313345.1 | - | 2 - 3 | 346 | 77 | ||
| BC030693.2 | - | 2 - 3 | 368 | 4827290 | 77 | |
Set Membership: |
|