| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009173.hg.1 |
ENST00000452788.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009173.hg.1 |
ENST00000452788.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009173.hg.1 |
NR_046632.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009173.hg.1 |
NR_046632.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00162873.hg.1 |
ENST00000452788.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00162873.hg.1 |
NR_046632.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00162873.hg.1 |
ENST00000452788.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00162873.hg.1 |
NR_046632.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00162873.hg.1 |
TC0X00009173.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00162874.hg.1 |
ENST00000452788.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00162874.hg.1 |
NR_046632.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00162874.hg.1 |
ENST00000452788.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00162874.hg.1 |
NR_046632.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00162874.hg.1 |
TC0X00009173.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0X00009173.hg.1 |
ENST00000452788.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009173.hg.1 |
ENST00000452788.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009173.hg.1 |
NR_046632.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009173.hg.1 |
NR_046632.1 |
Refseq |