Search
Search
| Gene Symbol: Csf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113530.1 | NP_001107002.1 | 5 - 6 | 954 | 66 | |
| NM_007778.4 | NP_031804.3 | 5 - 6 | 954 | 66 | ||
| GenBank mRNA | AK136808.1 | - | 5 - 6 | 728 | 66 | |
| AK138489.1 | - | 5 - 6 | 728 | 66 | ||
| AK154261.1 | - | 5 - 6 | 955 | 66 | ||
| AK154872.1 | - | 5 - 6 | 728 | 66 | ||
| AK160995.1 | - | 5 - 6 | 728 | 66 | ||
| AK166370.1 | - | 5 - 6 | 727 | 66 | ||
| AK170154.1 | - | 5 - 6 | 664 | 66 | ||
| BC066187.1 | - | 5 - 6 | 708 | 30106218 | 66 | |
| BC066200.1 | - | 5 - 6 | 708 | 30100524 | 66 | |
| BC066205.1 | - | 5 - 6 | 708 | 30100601 | 66 | |
| M21149.1 | - | 5 - 6 | 617 | 66 | ||
| M21952.1 | - | 5 - 6 | 707 | 66 | ||
| X05010.1 | - | 5 - 6 | 729 | 66 | ||
Set Membership: |
|