Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00433546_m1
          See other GCGR GE Assays ›
          Gene Symbol:
          Gcgr
          Gene Name:
          glucagon receptor
          Gene Aliases:
          GR
          Chromosome Location:
          Chr.11: 120529190 - 120538987 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00433546_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gcgr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
          RefSeq NM_008101.2 NP_032127.2 3 - 4 353 140
          GenBank mRNA AK004890.1 - 3 - 4 307 140
          AK144393.1 - 3 - 4 300 140
          BC012718.1 - 3 - 4 338 140
          BC031885.1 - 3 - 4 303 140
          BC057988.1 - 3 - 4 284 140
          L38613.1 - 2 - 3 263 140

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100002032.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gcgr

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          exocytosis
          signal transduction
          cell surface receptor signaling pathway
          G-protein coupled receptor signaling pathway
          adenylate cyclase-modulating G-protein coupled receptor signaling pathway
          adenylate cyclase-activating G-protein coupled receptor signaling pathway
          hormone-mediated signaling pathway
          glucose homeostasis
          response to starvation
          regulation of glycogen metabolic process
          cellular response to glucagon stimulus
          signal transducer activity
          transmembrane signaling receptor activity
          G-protein coupled receptor activity
          glucagon receptor activity
          peptide hormone binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100019575.mm.1 NM_008101.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100019575.mm.1 NM_008101.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100019575.mm.1 TC1100002032.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100019577.mm.1 NM_008101.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100019577.mm.1 NM_008101.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100019577.mm.1 TC1100002032.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100019576.mm.1 NM_008101.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100019576.mm.1 NM_008101.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100019576.mm.1 TC1100002032.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100019579.mm.1 NM_008101.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100019579.mm.1 NM_008101.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100019579.mm.1 TC1100002032.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100019572.mm.1 NM_008101.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100019572.mm.1 NM_008101.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100019572.mm.1 TC1100002032.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 621029 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-svw7g:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch