Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00434273_m1
          See other IL3RA GE Assays ›
          Gene Symbol:
          Il3ra
          Gene Name:
          interleukin 3 receptor, alpha chain
          Gene Aliases:
          CD123, CDw123, SUT-1
          Chromosome Location:
          Chr.14: 14346402 - 14355493 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00434273_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Il3ra
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008369.1 NP_032395.1 2 - 3 195 73
          GenBank mRNA BC049889.1 - 1 - 2 595 73
          BC077736.1 - 1 - 2 136 73
          X64534.1 - 2 - 3 195 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400000112.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Il3ra

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Process(es)

          cytokine receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2008181 X64534.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2008181 BC049889.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2008181 BC077736.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2008181 NM_008369.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 768480 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 814248 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 313123 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 156232 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 203284 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 150462 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P122107 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2008181 X64534.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2008181 BC049889.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2008181 BC077736.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2008181 NM_008369.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P432651 X64534.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P432651 BC049889.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P432651 FJ550346.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P432651 NM_008369.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29420290 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29420280 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29420291 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29420273 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29420278 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29420281 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29420268 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29420274 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29420266 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29420277 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29420267 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29420289 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29420286 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29420285 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29420272 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29420284 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29420276 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08034782 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P122107 - -
          Whole Mouse Genome G4122A A_51_P432651 X64534.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P432651 BC049889.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P432651 FJ550346.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P432651 NM_008369.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115035 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P122107 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11550459 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11550453 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11550478 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11550480 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11550456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29420303 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11550467 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11743831 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11550493 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29420300 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29420296 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29420295 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29420285 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29420303 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29420284 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29420266 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29420286 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29420305 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29420316 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29420312 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29420276 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29420293 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29420297 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29420299 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29420321 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29420317 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29420311 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29420290 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29420272 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29420292 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11550463 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29420309 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29420315 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29420314 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29420278 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11550459 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11550456 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11550462 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29420320 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29420313 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10412768 - -
          Affymetrix Mouse Gene 1.0 ST Array 10412770 - -
          Affymetrix Mouse Gene 1.0 ST Array 10412763 - -
          Affymetrix Mouse Gene 1.0 ST Array 10412762 - -
          Affymetrix Mouse Gene 1.0 ST Array 10412767 - -
          Affymetrix Mouse Gene 1.0 ST Array 10412765 - -
          Affymetrix Mouse Gene 1.0 ST Array 10412761 - -
          Affymetrix Mouse Gene 1.0 ST Array 10412766 - -
          Affymetrix Mouse Gene 1.0 ST Array 10412769 - -
          Affymetrix Mouse Gene 1.0 ST Array 10412764 - -
          Affymetrix HT MG-430 PM Array Plate 1419712_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10412760 - -
          Mouse Genome 430 2.0 Array 1419712_PM_at - -
          Mouse Genome 430 2.0 Array 1419712_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16188_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16188_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1400000900.mm.1 NM_008369.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400000900.mm.1 NM_008369.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400000900.mm.1 TC1400000112.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400000899.mm.1 NM_008369.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400000899.mm.1 NM_008369.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400000899.mm.1 TC1400000112.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400000901.mm.1 NM_008369.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400000901.mm.1 NM_008369.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400000901.mm.1 TC1400000112.mm.1 MTA TC
          Mouse Expression Set 430 1419712_PM_at - -
          Mouse Expression Set 430 1419712_at - -
          Murine Genome U74v2 Set 92955_at - -
          Murine Genome U74v2 Set 92955_PM_at - -
          Mouse Genome 430A 2.0 Array 1419712_PM_at - -
          Mouse Genome 430A 2.0 Array 1419712_at - -
          Affymetrix HT Mouse Genome 430A Array 1419712_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419712_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 446448 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6420722 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006420722 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2724449 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2724449 - -
          mouse-6 v1.1 (Illumina) scl46617.10.1_4-S - -
          Illumina Sentrix mouse-6.v1 bead array 6420722 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl46617.10.1_4-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6420722 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2916571 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2916571 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline