| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P237232 |
AK047512.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P237232 |
BC003803.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P237232 |
L34169.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P237232 |
NM_001173505.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P237232 |
NM_009379.3 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P237232 |
AK047512.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P237232 |
BC003803.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P237232 |
L34169.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P237232 |
NM_001173505.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P237232 |
NM_009379.3 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P119983 |
AK047512.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P119983 |
NM_001173505.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P119983 |
NM_009379.3 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
38779 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
284958 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
639981 |
AK035019.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
639981 |
BC051968.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
639981 |
NM_016743.2 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
618481 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
225050 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
67678 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
946146 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
458147 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
119415 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P126366 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P237232 |
AK047512.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P237232 |
BC003803.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P237232 |
L34169.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P237232 |
NM_001173505.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P237232 |
NM_009379.3 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P237232 |
AK047512.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P237232 |
BC003803.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P237232 |
L34169.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P237232 |
NM_001173505.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P237232 |
NM_009379.3 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P119983 |
AK047512.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P119983 |
NM_001173505.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P119983 |
NM_009379.3 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P385086 |
AK047512.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P385086 |
BC003803.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P385086 |
L34169.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P385086 |
NM_001173505.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P385086 |
NM_009379.3 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P237232 |
AK047512.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P237232 |
BC003803.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P237232 |
L34169.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P237232 |
NM_001173505.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P237232 |
NM_009379.3 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P237232 |
AK047512.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P237232 |
BC003803.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P237232 |
L34169.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P237232 |
NM_001173505.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P237232 |
NM_009379.3 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P431164 |
AK047512.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P431164 |
BC003803.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P431164 |
L34169.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P431164 |
NM_001173505.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P431164 |
NM_009379.3 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30589284 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30589275 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30589286 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30589277 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30589290 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30589282 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30589279 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30589287 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30589281 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30589288 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30589274 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30589289 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08527913 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P126366 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08527902 |
- |
- |
| Whole Mouse Genome G4122A |
A_52_P237232 |
AK047512.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P237232 |
BC003803.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P237232 |
L34169.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P237232 |
NM_001173505.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P237232 |
NM_009379.3 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P237232 |
AK047512.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P237232 |
BC003803.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P237232 |
L34169.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P237232 |
NM_001173505.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P237232 |
NM_009379.3 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P431164 |
AK047512.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P431164 |
BC003803.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P431164 |
L34169.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P431164 |
NM_001173505.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P431164 |
NM_009379.3 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P385086 |
AK047512.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P385086 |
BC003803.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P385086 |
L34169.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P385086 |
NM_001173505.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P385086 |
NM_009379.3 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P119983 |
AK047512.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P119983 |
NM_001173505.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P119983 |
NM_009379.3 |
mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P126366 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589321 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589243 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589173 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589166 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589163 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589172 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589342 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589184 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589311 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589206 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589169 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589171 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589241 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589284 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589335 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589235 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589155 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589168 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589178 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589181 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589266 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589192 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589217 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589327 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589253 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589278 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589259 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589338 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589161 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589208 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589185 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589296 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589156 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589249 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589162 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589269 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589180 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589230 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589214 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589225 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589244 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589337 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589158 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589286 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589213 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589195 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589240 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589268 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589242 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589256 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589152 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589250 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589202 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589310 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589226 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589302 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589222 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589175 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589265 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589323 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589331 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589282 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589211 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589247 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589281 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589287 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589329 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589215 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589288 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589160 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589316 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589300 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589294 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589219 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589325 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589340 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589290 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589186 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589317 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589305 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589307 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589237 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589279 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589309 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30589262 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12779251 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30589321 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30589332 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30589282 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30589323 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30589343 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30589305 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30589300 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30589314 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30589284 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30589316 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12751931 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30589338 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12779257 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30589335 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30589342 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30589307 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30589290 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30589286 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12480331 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30589302 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30589288 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30589309 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30589211 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13019161 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12779248 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12751926 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12480332 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13019162 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12779240 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30589329 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12790484 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30589278 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30589310 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12995906 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30589337 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12707529 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30589327 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30589213 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30589331 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12806550 |
- |
- |