| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100025833.mm.1 |
NM_009522.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100025833.mm.1 |
NM_009522.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100025833.mm.1 |
TC1100002848.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100025834.mm.1 |
NM_009522.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100025834.mm.1 |
NM_009522.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100025834.mm.1 |
TC1100002848.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100025831.mm.1 |
NM_009522.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100025831.mm.1 |
NM_009522.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100025831.mm.1 |
TC1100002848.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100025832.mm.1 |
NM_009522.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100025832.mm.1 |
NM_009522.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100025832.mm.1 |
TC1100002848.mm.1 |
MTA TC |