Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00438040_g1
          See other CASP14 GE Assays ›
          Gene Symbol:
          Casp14
          Gene Name:
          caspase 14
          Gene Aliases:
          MICE, mini-ICE
          Chromosome Location:
          Chr.10: 78711995 - 78718347 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00438040_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Casp14
          Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
          RefSeq NM_009809.5 NP_033939.1 2 - 3 255 67
          GenBank mRNA AF092997.1 - 1 - 2 100 67
          AJ007750.1 - 2 - 3 120 67
          AK081645.1 - 2 - 3 208 67
          BC101959.1 - 2 - 3 193 67
          BC101960.1 - 2 - 3 193 67
          BC103555.1 - 2 - 3 193 67
          BC103556.1 - 2 - 3 193 67
          BY134001.1 - 2 - 3 259 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000002460.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Casp14

          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          intrinsic apoptotic signaling pathway in response to DNA damage
          cell differentiation
          extrinsic apoptotic signaling pathway in absence of ligand
          cysteine-type endopeptidase activity
          peptidase activity
          cysteine-type peptidase activity
          hydrolase activity
          cysteine-type endopeptidase activity involved in apoptotic signaling pathway

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2064063 AJ007750.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2064063 AK081645.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2064063 AF092997.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2064063 NM_009809.5 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 518443 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 772262 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 343058 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 887657 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 165771 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P106342 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137474 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P139077 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2064063 AJ007750.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2064063 AK081645.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2064063 AF092997.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2064063 NM_009809.5 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P463159 AJ007750.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P463159 AK081645.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P463159 AF092997.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P463159 NM_009809.5 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275347 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275342 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275366 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275346 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275368 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275373 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275364 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275352 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275361 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275375 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275359 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275355 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275377 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275365 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275357 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275350 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275360 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275363 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275370 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27275348 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P139077 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P137474 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P106342 - -
          Whole Mouse Genome G4122A A_51_P463159 AJ007750.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P463159 AK081645.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P463159 AF092997.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P463159 NM_009809.5 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129930 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P139077 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P137474 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P106342 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08757878 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08757884 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08261254 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08757882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27275373 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275290 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275342 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275370 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275312 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275296 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275388 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275366 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275326 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275315 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275368 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275361 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275349 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275352 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08387080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275322 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08488087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08252079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08557964 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275382 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275291 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275355 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275383 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08429546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08261254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08252080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08261253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275336 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275373 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275292 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275339 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275380 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275324 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08757878 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08429550 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275319 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275372 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275378 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08761894 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275293 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275329 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275341 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275335 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275334 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08563098 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08252081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275344 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275346 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275350 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275318 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275359 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275328 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275371 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275320 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27275356 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10370525 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370529 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370523 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370528 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370524 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370526 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370527 - -
          Affymetrix HT MG-430 PM Array Plate 1443787_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1418748_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10370522 - -
          Mouse Genome 430 2.0 Array 1443787_PM_x_at - -
          Mouse Genome 430 2.0 Array 1418748_PM_at - -
          Mouse Genome 430 2.0 Array 1418748_at - -
          Mouse Genome 430 2.0 Array 1443787_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12365_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12365_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1000017932.mm.1 NM_009809.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000017932.mm.1 NM_009809.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1000017932.mm.1 TC1000002460.mm.1 MTA TC
          Mouse Expression Set 430 1418748_PM_at - -
          Mouse Expression Set 430 1418748_at - -
          Mouse Expression Set 430 1443787_PM_x_at - -
          Mouse Expression Set 430 1443787_x_at - -
          Murine Genome U74v2 Set 103837_at - -
          Murine Genome U74v2 Set 103837_PM_at - -
          Mouse Genome 430A 2.0 Array 1418748_PM_at - -
          Mouse Genome 430A 2.0 Array 1418748_at - -
          Affymetrix HT Mouse Genome 430A Array 1418748_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418748_at - -
          Affymetrix HT Mouse Genome 430B Array 1443787_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1443787_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 455803 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6510156 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006510156 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1241322 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1241322 - -
          mouse-6 v1.1 (Illumina) scl37766.7.1_51-S - -
          Illumina Sentrix mouse-6.v1 bead array 6510156 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl37766.7.1_51-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6510156 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1241322 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1241322 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch