Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › TaqMan® Gene Expression Assays
          • › Search Tool
          • › Search Results
          • › Mm00438875_m1
          See other FCGR2B GE Assays ›
          Gene Symbol:
          Fcgr2b
          Gene Name:
          Fc receptor, IgG, low affinity IIb
          Gene Aliases:
          AI528646, CD32, F630109E10Rik, Fc[g]RII, FcgRII, Fcgr2, Fcgr2a, Fcr-2, Fcr-3, Ly-17, Ly-m20, LyM-1, Lym-1, fcRII
          Chromosome Location:
          Chr.1: 170960558 - 170976071 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00438875_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fcgr2b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001077189.1 NP_001070657.1 1 - 2 174 77
          NM_010187.2 NP_034317.1 1 - 2 174 77
          GenBank mRNA AK080885.1 - 1 - 2 177 77
          AK089266.1 - 1 - 2 175 77
          AK150613.1 - 1 - 2 175 77
          AK155329.1 - 1 - 2 166 77
          AK170035.1 - 1 - 2 164 77
          AK171254.1 - 1 - 2 166 77
          BC011305.1 - 1 - 2 76
          BC019758.1 - 1 - 2 76
          BC038070.1 - 1 - 2 161 3993915 77
          M14216.1 - 1 - 2 132 77
          M14276.1 - 1 - 2 70 77
          M16367.1 - 3 - 4 415 77
          M17515.1 - 1 - 2 128 77
          U51629.1 - 1 - 2 82 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100003522.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the M14276.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fcgr2b

          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of type I hypersensitivity
          immune complex clearance
          negative regulation of immunoglobulin production
          negative regulation of inflammatory response to antigenic stimulus
          negative regulation of acute inflammatory response to antigenic stimulus
          negative regulation of humoral immune response mediated by circulating immunoglobulin
          receptor-mediated endocytosis
          phagocytosis, engulfment
          defense response
          immune response
          cell surface receptor signaling pathway
          response to bacterium
          immunoglobulin mediated immune response
          antigen processing and presentation of exogenous peptide antigen via MHC class II
          negative regulation of B cell proliferation
          endocytic recycling
          negative regulation of interleukin-10 production
          negative regulation of neutrophil apoptotic process
          intracellular signal transduction
          mast cell activation
          negative regulation of phagocytosis
          positive regulation of phagocytosis
          negative regulation of immune response
          cellular response to molecule of bacterial origin
          receptor signaling protein activity
          protein binding
          IgG binding
          immunoglobulin binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0100028826.mm.1 NM_001077189.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028826.mm.1 NM_001077189.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028826.mm.1 NM_010187.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028826.mm.1 NM_010187.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028826.mm.1 TC0100003522.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100028829.mm.1 NM_001077189.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028829.mm.1 NM_001077189.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028829.mm.1 NM_010187.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028829.mm.1 NM_010187.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028829.mm.1 TC0100003522.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 930585 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline