Search
Search

Prkca| Gene Symbol: Prkca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011101.3 | NP_035231.2 | 5 - 6 | 756 | 81 | |
| GenBank mRNA | AK147084.1 | - | 5 - 6 | 757 | 81 | |
| BC096493.1 | - | 5 - 6 | 550 | 30742571 | 81 | |
| M25811.1 | - | 5 - 6 | 790 | 81 | ||
| X52684.1 | - | 6 - 7 | 697 | 81 | ||
| X52685.1 | - | 6 - 7 | 697 | 81 | ||
Set Membership: |
|