Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00451387_m1
          See other KAT2B GE Assays ›
          Gene Symbol:
          Kat2b
          Gene Name:
          K(lysine) acetyltransferase 2B
          Gene Aliases:
          A930006P13Rik, AI461839, AW536563, Pcaf, p/CAF
          Chromosome Location:
          Chr.17: 53566853 - 53672721 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00451387_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kat2b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001190846.1 NP_001177775.1 14 - 15 1866 91
          NM_020005.4 NP_064389.2 14 - 15 2420 91
          GenBank mRNA AF254442.1 - 14 - 15 2061 91
          AK030070.1 - 14 - 15 1867 91
          AK156290.1 - 14 - 15 2420 91
          BC082581.1 - 14 - 15 2076 30653109 91
          BC138195.1 - 14 - 15 2088 8861214 91
          BC145896.1 - 14 - 15 2088 40131199 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000975.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus Mouse Androgen Panel Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kat2b

          Gene Ontology Categories:

          Function(s) Process(es)

          chromatin remodeling
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          protein acetylation
          cell cycle
          negative regulation of cell proliferation
          regulation of protein ADP-ribosylation
          positive regulation of neuron projection development
          histone acetylation
          N-terminal peptidyl-lysine acetylation
          internal peptidyl-lysine acetylation
          peptidyl-lysine acetylation
          cellular response to insulin stimulus
          positive regulation of chromatin binding
          positive regulation of gluconeogenesis by positive regulation of transcription from RNA polymerase II promoter
          histone H3 acetylation
          histone H3-K9 acetylation
          negative regulation of cyclin-dependent protein serine/threonine kinase activity
          positive regulation of transcription, DNA-templated
          positive regulation of vasodilation
          positive regulation of transcription from RNA polymerase II promoter
          rhythmic process
          positive regulation of histone H3-K14 acetylation
          positive regulation of histone H3-K9 acetylation
          RNA polymerase II regulatory region sequence-specific DNA binding
          chromatin binding
          transcription cofactor activity
          transcription coactivator activity
          histone acetyltransferase activity
          lysine N-acetyltransferase activity, acting on acetyl phosphate as donor
          cyclin-dependent protein serine/threonine kinase inhibitor activity
          protein binding
          N-acetyltransferase activity
          transcription factor binding
          acetyltransferase activity
          transferase activity
          transferase activity, transferring acyl groups
          protein kinase binding
          protein complex binding
          histone acetyltransferase binding
          histone deacetylase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1700009336.mm.1 NM_001190846.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009336.mm.1 NM_001190846.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009336.mm.1 NM_020005.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009336.mm.1 NM_020005.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009336.mm.1 TC1700000975.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700009337.mm.1 NM_001190846.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009337.mm.1 NM_001190846.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009337.mm.1 NM_020005.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009337.mm.1 NM_020005.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009337.mm.1 TC1700000975.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700009334.mm.1 NM_001190846.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009334.mm.1 NM_001190846.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009334.mm.1 NM_020005.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009334.mm.1 NM_020005.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009334.mm.1 TC1700000975.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700009335.mm.1 NM_001190846.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009335.mm.1 NM_001190846.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009335.mm.1 NM_020005.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009335.mm.1 NM_020005.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009335.mm.1 TC1700000975.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700009339.mm.1 NM_001190846.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009339.mm.1 NM_001190846.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009339.mm.1 NM_020005.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009339.mm.1 NM_020005.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009339.mm.1 TC1700000975.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700009340.mm.1 NM_001190846.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009340.mm.1 NM_001190846.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009340.mm.1 NM_020005.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009340.mm.1 NM_020005.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009340.mm.1 TC1700000975.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline