Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00458118_m1
          See other DACT1 GE Assays ›
          Gene Symbol:
          Dact1
          Gene Name:
          dapper homolog 1, antagonist of beta-catenin (xenopus)
          Gene Aliases:
          4921528D17Rik, AI115603, DAPPER, DAPPER1, FRODO, Frd1, Frodo1, MDpr1, MTNG3, THYEX3
          Chromosome Location:
          Chr.12: 71309884 - 71320107 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00458118_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dact1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021532.4 NP_067507.2 3 - 4 718 86
          GenBank mRNA AF251078.1 - 1 - 2 54 86
          AF488775.1 - 3 - 4 618 86
          AK077691.1 - 3 - 4 335 86
          AK199966.1 - 1 - 2 44 86
          AY208970.1 - 3 - 4 570 86
          BC145833.1 - 3 - 4 668 40131136 86

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AF251078.1, AK077691.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus Mouse Wnt Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dact1

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          gastrulation with mouth forming second
          multicellular organism development
          nervous system development
          Wnt signaling pathway
          positive regulation of Wnt signaling pathway
          negative regulation of Wnt signaling pathway
          regulation of protein stability
          regulation of catenin import into nucleus
          positive regulation of catenin import into nucleus
          positive regulation of fat cell differentiation
          positive regulation of protein catabolic process
          negative regulation of JNK cascade
          embryonic morphogenesis
          embryonic hindgut morphogenesis
          dendrite morphogenesis
          synapse organization
          canonical Wnt signaling pathway
          negative regulation of canonical Wnt signaling pathway
          regulation of nodal signaling pathway
          regulation of Wnt signaling pathway, planar cell polarity pathway
          negative regulation of G1/S transition of mitotic cell cycle
          protein kinase C binding
          protein binding
          beta-catenin binding
          protein kinase A binding
          delta-catenin binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1200004189.mm.1 NM_001190466.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200004189.mm.1 NM_001190466.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200004189.mm.1 NM_021532.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200004189.mm.1 NM_021532.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1200004189.mm.1 TC1200000608.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200004190.mm.1 NM_001190466.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200004190.mm.1 NM_001190466.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200004190.mm.1 NM_021532.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200004190.mm.1 NM_021532.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1200004190.mm.1 TC1200000608.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200004191.mm.1 NM_001190466.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200004191.mm.1 NM_001190466.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200004191.mm.1 NM_021532.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200004191.mm.1 NM_021532.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1200004191.mm.1 TC1200000608.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 379520 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline