Search
Search

Unc5a| Gene Symbol: Unc5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153131.3 | NP_694771.1 | 14 - 15 | 2586 | 71 | |
| GenBank mRNA | AJ487852.1 | - | 14 - 15 | 2587 | 71 | |
| AK122575.1 | - | 10 - 11 | 2908 | 71 | ||
| BC058084.1 | - | 13 - 14 | 2439 | 6813463 | 71 | |
Set Membership: |
|