Search
Search

Ttc39a| Gene Symbol: Ttc39a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145948.1 | NP_001139420.1 | 10 - 11 | 894 | 58 | |
| NM_153392.2 | NP_700441.2 | 10 - 11 | 903 | 58 | ||
| GenBank mRNA | AK048659.1 | - | 10 - 11 | 904 | 58 | |
| AK076641.1 | - | 10 - 11 | 856 | 58 | ||
| AK076720.1 | - | 10 - 11 | 895 | 58 | ||
| AK076772.1 | - | 10 - 11 | 857 | 58 | ||
| AK172951.1 | - | 10 - 11 | 826 | 58 | ||
| BC023883.1 | - | 10 - 11 | 902 | 5150841 | 58 | |
| BC026660.1 | - | 10 - 11 | 844 | 4988900 | 58 | |
Set Membership: |
|