Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
New Products
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Mm00464475_m1
See other DMD GE Assays ›
Gene Symbol:
Dmd
Gene Name:
dystrophin, muscular dystrophy
Gene Aliases:
DXSmh7, DXSmh9, Dp427, Dp71, dys, mdx, pke
Chromosome Location:
Chr.X: 82310713 - 85205050 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID
Mm00464475_m1
Size
FAM (900 nM Primer) | XS: 75 rxns
FAM (900 nM Primer) | S: 250 rxns
FAM (900 nM Primer) | M: 750 rxns
FAM (900 nM Primer) | L: 2,900 rxns
FAM PL (350 nM Primer Limited) | S: 360 rxns
FAM PL (350 nM Primer Limited) | M: 750 rxns
FAM PL (350 nM Primer Limited) | L: 2,900 rxns
VIC (900 nM Primer) | S: 360 rxns
VIC (900 nM Primer) | M: 750 rxns
VIC (900 nM Primer) | L: 2,900 rxns
VIC PL (150 nM Primer Limited) | S: 360 rxns
VIC PL (150 nM Primer Limited) | M: 750 rxns
VIC PL (150 nM Primer Limited) | L: 2,900 rxns
ABY PL (350 nM Primer Limited) | S: 360 rxns
ABY PL (350 nM Primer Limited) | M: 750 rxns
ABY PL (350 nM Primer Limited) | L: 2,900 rxns
JUN PL (350 nM Primer Limited) | S: 360 rxns
JUN PL (350 nM Primer Limited) | M: 750 rxns
JUN PL (350 nM Primer Limited) | L: 2,900 rxns
CYN5 PL (350 nM Primer Limited) | S: 360 rxns
CYN5 PL (350 nM Primer Limited) | M: 750 rxns
CYN5 PL (350 nM Primer Limited) | L: 2,900 rxns
Availability
Inventoried
Catalog #
4331182
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Dmd
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
Amplicon Length
RefSeq
NM_007868.5
NP_031894.1
16 - 17
2247
83
GenBank mRNA
AK044536.1
-
16 - 17
2047
83
M68859.1
-
16 - 17
2220
83
Back To Top
More Information
Updated Mapping Information
This assay targets the same location as MTA transcript cluster(s) TC0X00000808.mm.1
Set Membership:
Ambion siRNA available
Amplicon length between 71 and 85
Mus musculus
1700
Inventoried
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene Dmd
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of cell-matrix adhesion
regulation of heart rate
regulation of transcription, DNA-templated
muscle organ development
skeletal muscle tissue development
establishment of blood-nerve barrier
regulation of gene expression
regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
positive regulation of neuron projection development
regulation of skeletal muscle contraction by regulation of release of sequestered calcium ion
regulation of skeletal muscle contraction
myotube cell development
muscle attachment
olfactory nerve structural organization
negative regulation of peptidyl-serine phosphorylation
cellular protein localization
response to muscle stretch
regulation of membrane potential
peptide biosynthetic process
cellular protein complex assembly
neurotransmitter receptor metabolic process
positive regulation of neuron differentiation
muscle cell cellular homeostasis
muscle fiber development
nucleus localization
cardiac muscle contraction
regulation of ryanodine-sensitive calcium-release channel activity
establishment of glial blood-brain barrier
negative regulation of ERK1 and ERK2 cascade
cardiac muscle cell action potential
regulation of cellular response to growth factor stimulus
regulation of voltage-gated calcium channel activity
negative regulation of peptidyl-cysteine S-nitrosylation
positive regulation of sodium ion transmembrane transporter activity
dystroglycan binding
actin binding
structural constituent of cytoskeleton
protein binding
zinc ion binding
structural constituent of muscle
myosin binding
vinculin binding
metal ion binding
nitric-oxide synthase binding
MicroArray Probes:
Affymetrix
ABI
Array Name
Probe IDs
Targets
Target Type
Affymetrix Mouse Clariom S Array
PSR0X00005388.mm.1
NM_007868.6
Refseq
Affymetrix Mouse Clariom S Array
PSR0X00005388.mm.1
TC0X00000808.mm.1
MTA TC
Array Name
Probe IDs
Targets
Target Type
ABI Mouse Genome Survey Microarray
813948
-
-
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-5db85fbf84-frpqk:80/100.66.72.150:80.
git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.60.1-2026-secondary-format-patch