Search
Search

Tbl1xr1| Gene Symbol: Tbl1xr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030732.3 | NP_109657.2 | 11 - 12 | 1459 | 89 | |
| GenBank mRNA | AF268195.1 | - | 11 - 12 | 1240 | 89 | |
| AK029595.1 | - | 11 - 12 | 1258 | 89 | ||
| AK033347.1 | - | 11 - 12 | 1329 | 89 | ||
| AK036064.1 | - | 12 - 13 | 1570 | 89 | ||
| AK136255.1 | - | 11 - 12 | 1281 | 89 | ||
| BC018512.1 | - | 1 - 2 | 105 | 89 | ||
Set Membership: |
|