| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700001622.mm.1 |
NM_001277988.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700001622.mm.1 |
NM_001277989.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700001622.mm.1 |
NM_008627.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700001622.mm.1 |
NR_102730.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700001622.mm.1 |
TC0700000223.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700001627.mm.1 |
NM_001277988.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700001627.mm.1 |
NM_001277989.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700001627.mm.1 |
NM_008627.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700001627.mm.1 |
NR_102730.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700001627.mm.1 |
TC0700000223.mm.1 |
MTA TC |