Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00486522_s1
          See other TSSK2 GE Assays ›
          Gene Symbol:
          Tssk2
          Gene Name:
          testis-specific serine kinase 2
          Gene Aliases:
          DGS-G, SPOGA2, Stk22b, Tsk2
          Chromosome Location:
          Chr.16: 17898637 - 17900024 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00486522_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tssk2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009436.2 NP_033462.2 1 928 117
          GenBank mRNA AF019926.1 - 1 854 117
          BC049618.1 - 1 911 117
          BC061175.1 - 1 927 6772604 117

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600000228.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tssk2

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          multicellular organism development
          spermatogenesis
          spermatid development
          phosphorylation
          cell differentiation
          intracellular signal transduction
          protein autophosphorylation
          nucleotide binding
          magnesium ion binding
          protein kinase activity
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30576408 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P402044 BC049618.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P402044 NM_009436.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P402044 AF019926.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P402044 BC049618.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P402044 NM_009436.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P402044 AF019926.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 960676 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 21860 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P176285 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P402044 BC049618.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P402044 NM_009436.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P402044 AF019926.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P402044 BC049618.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P402044 NM_009436.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P402044 AF019926.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30576394 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30576391 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30576382 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30576404 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30576415 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30576412 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30576402 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30576389 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30576384 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30576379 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30576419 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30576396 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30576398 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30576399 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30576418 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30576407 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30576386 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30576422 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30576390 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30576410 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30576416 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30576393 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30576401 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30576414 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30576420 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P176285 - -
          Whole Mouse Genome G4122A A_51_P402044 BC049618.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P402044 NM_009436.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P402044 AF019926.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P402044 BC049618.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P402044 NM_009436.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P402044 AF019926.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103108 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P176285 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12541095 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12541102 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12541090 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12541091 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12541126 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12541088 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30576408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30576379 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30576419 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30576398 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30576407 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12541091 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30576391 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30576416 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30576379 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12541088 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12541117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30576414 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30576384 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30576386 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30576396 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12541112 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30576401 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12541087 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30576410 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30576412 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30576390 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30576389 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12541129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12541111 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12541074 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10434127 - -
          Affymetrix Mouse Gene 1.0 ST Array 10434125 - -
          Affymetrix Mouse Gene 1.0 ST Array 10434126 - -
          Affymetrix HT MG-430 PM Array Plate 1449489_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10434124 - -
          Mouse Genome 430 2.0 Array 1449489_at - -
          Mouse Genome 430 2.0 Array 1449489_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22115_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22115_at - -
          Affymetrix Mouse Clariom S Array PSR1600002043.mm.1 NM_009436.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600002043.mm.1 NM_009436.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600002043.mm.1 TC1600000228.mm.1 MTA TC
          Mouse Expression Set 430 1449489_PM_at - -
          Mouse Expression Set 430 1449489_at - -
          Murine Genome U74v2 Set 102846_PM_at - -
          Murine Genome U74v2 Set 102846_at - -
          Mouse Genome 430A 2.0 Array 1449489_at - -
          Mouse Genome 430A 2.0 Array 1449489_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449489_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449489_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 325194 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5220324 - -
          Illumina mouse-6 v1.0 expression beadchip 5220324 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005220324 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2774952 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2774952 - -
          mouse-6 v1.1 (Illumina) scl022115.1_229-S - -
          Illumina Sentrix mouse-6.v1 bead array 5220324 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl022115.1_229-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5220324 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2774950 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2774950 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline