Search
Search

Gata2| Gene Symbol: Gata2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008090.5 | NP_032116.4 | 5 - 6 | 1402 | 66 | |
| GenBank mRNA | AB000096.2 | - | 5 - 6 | 1294 | 66 | |
| AK004551.1 | - | 5 - 6 | 1399 | 66 | ||
| AK004675.1 | - | 5 - 6 | 1731 | 66 | ||
| AK154792.1 | - | 5 - 6 | 1400 | 66 | ||
| AK154981.1 | - | 5 - 6 | 1403 | 66 | ||
| BC107009.2 | - | 5 - 6 | 1306 | 40047573 | 66 | |
| BC107010.1 | - | 4 - 5 | 1079 | 40047574 | 66 | |
Set Membership: |
|