Search
Search

Slc17a6| Gene Symbol: Slc17a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080853.3 | NP_543129.3 | 3 - 4 | 1261 | 83 | |
| GenBank mRNA | AF324864.1 | - | 3 - 4 | 1094 | 83 | |
| AK043753.1 | - | 3 - 4 | 802 | 83 | ||
| BC038375.1 | - | 3 - 4 | 640 | 5357143 | 83 | |
Set Membership: |
|