Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00501176_m1
          See other PRL7A1 GE Assays ›
          Gene Symbol:
          Prl7a1
          Gene Name:
          prolactin family 7, subfamily a, member 1
          Gene Aliases:
          AA407021, AA409971, AA410116, PLP-E, PLP-G, Prlpe, Prlpg
          Chromosome Location:
          Chr.13: 27633364 - 27642493 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00501176_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prl7a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164058.1 NP_001157530.1 6 - 7 621 75
          NM_008930.2 NP_032956.1 6 - 7 618 75
          GenBank mRNA AF011381.1 - 6 - 7 610 75
          AF020525.1 - 6 - 7 592 75
          AK141309.1 - 6 - 7 622 75
          AK146050.1 - 6 - 7 621 75
          BC051671.1 - 6 - 7 603 30018275 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300001695.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prl7a1

          Panther Classification:

          Molecular Function -

          peptide hormone

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          hormone activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2274782 AK028371.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2274782 AK141309.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2274782 BB611867.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2274782 BC051671.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2274782 NM_001164058.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 472606 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 272409 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 74619 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 142243 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 231794 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 57777 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P110975 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2274782 AK028371.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2274782 AK141309.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2274782 BB611867.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2274782 BC051671.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2274782 NM_001164058.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28903024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28902996 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28903013 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28902994 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28903002 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28903017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28903009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28903005 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28903020 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28902992 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28903021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28902998 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28903022 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28903028 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28903011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28902999 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28903015 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28903000 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28903008 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28902991 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28903019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28902990 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28903007 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28903004 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28903003 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28903026 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P110975 - -
          Whole Mouse Genome G4122A A_51_P211695 AK028371.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P211695 AK141309.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P211695 AK146050.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P211695 BB611867.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P211695 BC051671.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124557 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P110975 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10641023 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10935555 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10501835 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10501829 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10641022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28903000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28903019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28903004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28903028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28903011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28903007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28903032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28903024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28903021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28903031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28903030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28903013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28903003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28903002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28903023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28903009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28903026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28902917 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28902955 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28902986 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10641021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28903026 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10962453 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28903032 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28903006 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28903000 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28902979 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28903015 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28902972 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28902974 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28902983 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28902992 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28902994 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28903018 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28903031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28902958 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28903021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28903002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28902951 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28902964 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28903011 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28903023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28902970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28903017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28903013 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28902948 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28902985 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28902990 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10962457 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28903019 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28902982 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28902965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28902961 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28903007 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28902957 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28902975 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28903030 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28902996 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28903020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28902978 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28903009 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28902987 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28903028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10641023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28902949 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28903024 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28902950 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28902973 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11037513 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28902967 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28902999 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28902998 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10408421 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408425 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408424 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408426 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408422 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408423 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408427 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408428 - -
          Affymetrix HT MG-430 PM Array Plate 1441873_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1441874_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1449529_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10408420 - -
          Mouse Genome 430 2.0 Array 1449529_s_at - -
          Mouse Genome 430 2.0 Array 1441874_PM_x_at - -
          Mouse Genome 430 2.0 Array 1441874_x_at - -
          Mouse Genome 430 2.0 Array 1441873_at - -
          Mouse Genome 430 2.0 Array 1441873_PM_at - -
          Mouse Genome 430 2.0 Array 1449529_PM_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19113_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19113_at - -
          Affymetrix Mouse Clariom S Array PSR1300010546.mm.1 NM_001164058.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300010546.mm.1 NM_001164058.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300010546.mm.1 NM_008930.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300010546.mm.1 NM_008930.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300010546.mm.1 TC1300001695.mm.1 MTA TC
          Mouse Expression Set 430 1449529_s_at - -
          Mouse Expression Set 430 1449529_PM_s_at - -
          Mouse Expression Set 430 1441873_at - -
          Mouse Expression Set 430 1441874_PM_x_at - -
          Mouse Expression Set 430 1441873_PM_at - -
          Mouse Expression Set 430 1441874_x_at - -
          Murine Genome U74v2 Set 169538_i_at - -
          Murine Genome U74v2 Set 169538_PM_i_at - -
          Mouse Genome 430A 2.0 Array 1449529_s_at - -
          Mouse Genome 430A 2.0 Array 1449529_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1449529_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1449529_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1441874_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1441873_at - -
          Affymetrix HT Mouse Genome 430B Array 1441873_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1441874_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5130022 - -
          Illumina mouse-6 v1.0 expression beadchip 104150341 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005130022 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2606903 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1240343 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2606903 - -
          mouse-6 v1.1 (Illumina) scl44167.7.1_220-S - -
          Illumina Sentrix mouse-6.v1 bead array 5130022 - -
          Illumina Sentrix mouse-6.v1 bead array 104150341 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl44167.7.1_220-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104150341 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5130022 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2606903 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1240343 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2606903 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline