Search
Search

Gsdmd| Gene Symbol: Gsdmd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026960.4 | NP_081236.1 | 9 - 10 | 1297 | 83 | |
| GenBank mRNA | AK007710.1 | - | 9 - 10 | 1298 | 83 | |
| AK088747.1 | - | 9 - 10 | 1309 | 83 | ||
| AK165858.1 | - | 9 - 10 | 1298 | 83 | ||
| AK171294.1 | - | 9 - 10 | 1299 | 83 | ||
| BC029813.1 | - | 9 - 10 | 1250 | 4194837 | 83 | |
Set Membership: |
|