Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00511068_s1
          See other DDX28 GE Assays ›
          Gene Symbol:
          Ddx28
          Gene Name:
          DEAD (Asp-Glu-Ala-Asp) box polypeptide 28
          Gene Aliases:
          2410004K13Rik, AI449652, Mddx28
          Chromosome Location:
          Chr.8: 106009616 - 106011486 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00511068_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ddx28
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028038.3 NP_082314.2 1 308 90
          GenBank mRNA AK010396.1 - 1 256 90
          AK144945.1 - 1 702 90
          AK163480.1 - 1 261 90
          BC058568.1 - 1 268 90
          BC120556.1 - 1 308 40129479 90
          BC120582.1 - 1 308 40129505 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800002848.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ddx28

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          RNA secondary structure unwinding
          nucleotide binding
          nucleic acid binding
          RNA binding
          ATP-dependent RNA helicase activity
          helicase activity
          ATP binding
          hydrolase activity
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137275 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137273 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137276 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137271 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137272 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137277 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P219325 BC120582.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P219325 AK163480.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P219325 BC058568.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P219325 BC120556.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P219325 AK010396.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P219325 BC120582.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P219325 AK163480.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P219325 BC058568.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P219325 BC120556.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P219325 AK010396.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 485624 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 128775 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P103238 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P219325 BC120582.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P219325 AK163480.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P219325 BC058568.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P219325 BC120556.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P219325 AK010396.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P219325 BC120582.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P219325 AK163480.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P219325 BC058568.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P219325 BC120556.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P219325 AK010396.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137291 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137293 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137282 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137279 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137257 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137283 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137276 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137289 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137264 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137262 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137266 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137273 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137281 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137269 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137288 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137268 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137297 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137260 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137285 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137292 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137286 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137270 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137294 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137299 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137258 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P103238 - -
          Whole Mouse Genome G4122A A_51_P219325 BC120582.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P219325 AK163480.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P219325 BC058568.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P219325 BC120556.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P219325 AK010396.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P219325 BC120582.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P219325 AK163480.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P219325 BC058568.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P219325 BC120556.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P219325 AK010396.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139617 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P103238 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06738724 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137272 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137273 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137275 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137276 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137271 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137268 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07060440 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137272 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137255 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137274 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137250 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137268 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06738719 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06738718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137269 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137262 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06738699 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10581454 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581451 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581452 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581453 - -
          Affymetrix HT MG-430 PM Array Plate 1428702_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10581450 - -
          Mouse Genome 430 2.0 Array 1428702_at - -
          Mouse Genome 430 2.0 Array 1428702_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71986_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71986_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0800021984.mm.1 NM_028038.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021984.mm.1 NM_028038.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021984.mm.1 TC0800002848.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800021985.mm.1 NM_028038.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021985.mm.1 NM_028038.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021985.mm.1 TC0800002848.mm.1 MTA TC
          Mouse Expression Set 430 1428702_at - -
          Mouse Expression Set 430 1428702_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1428702_at - -
          Affymetrix HT Mouse Genome 430B Array 1428702_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 316375 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1170603 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001170603 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1259277 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1259277 - -
          mouse-6 v1.1 (Illumina) scl34380.1_1-S - -
          Illumina Sentrix mouse-6.v1 bead array 1170603 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl34380.1_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1170603 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1259277 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1259277 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline