Search
Search

Tmem43| Gene Symbol: Tmem43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028766.2 | NP_083042.1 | 10 - 11 | 1015 | 71 | |
| GenBank mRNA | AK004778.1 | - | 10 - 11 | 1016 | 71 | |
| AK037165.1 | - | 10 - 11 | 986 | 71 | ||
| AK050173.1 | - | 10 - 11 | 1055 | 71 | ||
| AK151693.1 | - | 10 - 11 | 1008 | 71 | ||
| AK151766.1 | - | 10 - 11 | 1033 | 71 | ||
| AK152078.1 | - | 10 - 11 | 1008 | 71 | ||
| AK168559.1 | - | 10 - 11 | 1023 | 71 | ||
| BC005521.1 | - | 10 - 11 | 981 | 71 | ||
| BC024933.1 | - | 10 - 11 | 1002 | 3990353 | 71 | |
Set Membership: |
|