Search
Search

Tspan17| Gene Symbol: Tspan17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028841.3 | NP_083117.2 | 6 - 7 | 821 | 88 | |
| GenBank mRNA | AI019499.1 | - | 3 - 4 | 292 | 88 | |
| AK008761.1 | - | 6 - 7 | 88 | |||
| AK166272.1 | - | 6 - 7 | 824 | 88 | ||
| AK171691.1 | - | 6 - 7 | 770 | 88 | ||
| BC010346.1 | - | 6 - 7 | 766 | 2811935 | 88 | |
| CD740983.1 | - | 3 - 4 | 88 | |||
Set Membership: |
|