Search
Search
| Gene Symbol: Ctsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007798.3 | NP_031824.1 | 5 - 6 | 577 | 63 | |
| GenBank mRNA | AK002456.1 | - | 5 - 6 | 526 | 63 | |
| AK083393.1 | - | 5 - 6 | 577 | 63 | ||
| AK147192.1 | - | 5 - 6 | 524 | 63 | ||
| AK149884.1 | - | 5 - 6 | 526 | 63 | ||
| AK151790.1 | - | 5 - 6 | 524 | 63 | ||
| AK153047.1 | - | 5 - 6 | 523 | 63 | ||
| AK159992.1 | - | 5 - 6 | 562 | 63 | ||
| AK167361.1 | - | 5 - 6 | 558 | 63 | ||
| AK170959.1 | - | 5 - 6 | 550 | 63 | ||
| AK192561.1 | - | 1 - 2 | 59 | 63 | ||
| AK197104.1 | - | 1 - 2 | 59 | 63 | ||
| AK198001.1 | - | 1 - 2 | 59 | 63 | ||
| AK198619.1 | - | 1 - 2 | 59 | 63 | ||
| AK198709.1 | - | 1 - 2 | 59 | 63 | ||
| AK201305.1 | - | 1 - 2 | 59 | 63 | ||
| AK201756.1 | - | 1 - 2 | 59 | 63 | ||
| AK202640.1 | - | 1 - 2 | 59 | 63 | ||
| AK203701.1 | - | 1 - 2 | 63 | |||
| AK205609.1 | - | 1 - 2 | 59 | 63 | ||
| AK210127.1 | - | 1 - 2 | 59 | 63 | ||
| AK213062.1 | - | 1 - 2 | 59 | 63 | ||
| AK213791.1 | - | 1 - 2 | 59 | 63 | ||
| BC006656.1 | - | 5 - 6 | 568 | 3500700 | 63 | |
| S69034.1 | - | 5 - 6 | 573 | 63 | ||
Set Membership: |
|