Search
Search

Stap2| Gene Symbol: Stap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145934.2 | NP_666046.1 | 1 - 2 | 176 | 71 | |
| NR_102313.1 | 1 - 2 | 176 | 71 | |||
| GenBank mRNA | AK050131.1 | - | 1 - 2 | 156 | 71 | |
| AK050172.1 | - | 1 - 2 | 144 | 71 | ||
| BC026642.1 | - | 1 - 2 | 150 | 4986113 | 71 | |
Set Membership: |
|