Search
Search

Lta4h| Gene Symbol: Lta4h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008517.2 | NP_032543.2 | 10 - 11 | 1060 | 80 | |
| GenBank mRNA | AK134931.1 | - | 10 - 11 | 1046 | 80 | |
| BC021417.1 | - | 10 - 11 | 1054 | 5004113 | 80 | |
| M63848.1 | - | 1026 | 80 | |||
Set Membership: |
|