Search
Search

Galnt13| Gene Symbol: Galnt13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173030.2 | NP_766618.2 | 8 - 9 | 1488 | 70 | |
| GenBank mRNA | AB082928.1 | - | 6 - 7 | 976 | 70 | |
| AK030726.1 | - | 7 - 8 | 1420 | 70 | ||
| AK038387.1 | - | 8 - 9 | 1489 | 70 | ||
| AK045417.1 | - | 8 - 9 | 1489 | 70 | ||
| BC131651.1 | - | 8 - 9 | 1167 | 40110517 | 70 | |
| BC131652.1 | - | 7 - 8 | 1198 | 40110518 | 70 | |
Set Membership: |
|