Search
Search

Galk2| Gene Symbol: Galk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291002.1 | NP_001277931.1 | 8 - 9 | 1080 | 64 | |
| NM_175154.3 | NP_780363.1 | 8 - 9 | 1186 | 64 | ||
| GenBank mRNA | AK012757.1 | - | 1 - 2 | 42 | 64 | |
| AK082521.1 | - | 8 - 9 | 1077 | 64 | ||
| AK152851.1 | - | 6 - 7 | 769 | 64 | ||
| AK154762.1 | - | 8 - 9 | 1035 | 64 | ||
| BC079843.1 | - | 8 - 9 | 1050 | 5690056 | 64 | |
Set Membership: |
|