Search
Search
| Gene Symbol: Ano6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253813.1 | NP_001240742.1 | 14 - 15 | 1823 | 73 | |
| NM_175344.4 | NP_780553.2 | 13 - 14 | 1760 | 73 | ||
| GenBank mRNA | AK089300.1 | - | 14 - 15 | 1896 | 73 | |
| AK159411.1 | - | 14 - 15 | 1772 | 73 | ||
| BC060732.1 | - | 13 - 14 | 1760 | 6409332 | 73 | |
Set Membership: |
|