Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00617657_m1
          See other PRSS33 GE Assays ›
          Gene Symbol:
          Prss33
          Gene Name:
          protease, serine 33
          Gene Aliases:
          Eos, mT6, tryptase-6
          Chromosome Location:
          Chr.17: 23833338 - 23836126 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00617657_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prss33
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001081399.2 NP_001074868.2 1 - 2 92 77
          GenBank mRNA AY262280.1 - 1 - 2 68 77
          BC101950.1 - 1 - 2 68 40044230 77
          BC103542.1 - 1 - 2 68 40044219 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700001607.mm.1

          Set Membership:

          Amplicon length between 71 and 85 Protease Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          serine-type endopeptidase activity
          peptidase activity
          serine-type peptidase activity
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P342654 NM_001081399.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P342654 AY262280.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P342654 BC101950.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P342654 BC103541.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P342654 BC103542.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P342654 NM_001081399.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P342654 AY262280.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P342654 BC101950.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P342654 BC103541.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P342654 BC103542.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 125627 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P150011 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P342654 NM_001081399.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P342654 AY262280.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P342654 BC101950.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P342654 BC103541.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P342654 BC103542.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P342654 NM_001081399.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P342654 AY262280.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P342654 BC101950.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P342654 BC103541.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P342654 BC103542.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P150011 - -
          Whole Mouse Genome G4122A A_52_P342654 NM_001081399.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P342654 AY262280.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P342654 BC101950.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P342654 BC103541.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P342654 BC103542.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P342654 NM_001081399.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P342654 AY262280.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P342654 BC101950.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P342654 BC103541.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P342654 BC103542.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P150011 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13127261 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13127239 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13127270 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13127231 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13127236 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13127244 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13127237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093906 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093953 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093946 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093941 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093934 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093936 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093952 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093938 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093947 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093958 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093955 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13127228 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093948 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093960 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13127258 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093943 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10448388 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448387 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448389 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448385 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448386 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448384 - -
          Affymetrix Mouse Gene 1.1 ST Array 10448383 - -
          Affymetrix Mouse Clariom S Array PSR1700014089.mm.1 NM_001081399.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014089.mm.1 NM_001081399.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014089.mm.1 TC1700001607.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700014083.mm.1 NM_001081399.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014083.mm.1 NM_001081399.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014083.mm.1 TC1700001607.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 428610 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1500450 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1231590 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1231590 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1231590 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline